Merck
  • Synthesis of new vitamin K analogues as steroid and xenobiotic receptor (SXR) agonists: insights into the biological role of the side chain part of vitamin K.
Synthesis of new vitamin K analogues as steroid and xenobiotic receptor (SXR) agonists: insights into the biological role of the side chain part of vitamin K.
Journal of medicinal chemistry (2011-05-31)
Yoshitomo Suhara, Masato Watanabe, Sayaka Motoyoshi, Kimie Nakagawa, Akimori Wada, Kazuyoshi Takeda, Kazuhiko Takahashi, Hiroaki Tokiwa, Toshio Okano
ABSTRACT
Vitamin K(2) has been demonstrated to induce gene expression related to bone formation through a nuclear steroid and xenobiotic receptor (SXR). We synthesized new vitamin K analogues with the same isoprene side chains symmetrically introduced at the 2 and 3 positions of 1,4-naphthoquinone and evaluated the transcriptional activity of the target gene. The transcriptional activity was related to the length of the side chain which allowed optimal interaction with ligand-binding domain of SXR.
Sigma-Aldrich
Vitamin K2
Sigma-Aldrich
Vitamin K1, BioXtra, ≥99.0% (sum of isomers, HPLC), mixture of isomers
Sigma-Aldrich
Vitamin K1, viscous liquid
Supelco
Phylloquinone (K1), analytical standard
Sigma-Aldrich
Rifampicin, suitable for plant cell culture, BioReagent, ≥95% (HPLC), powder or crystals
Sigma-Aldrich
Rifampicin, ≥95% (HPLC), powder or crystals