Skip to Content
Merck
All Photos(4)

Key Documents

205621

Sigma-Aldrich

Diethylene glycol dibutyl ether

≥99%

Synonym(s):

2-Butoxyethyl ether, Bis(2-butoxyethyl) ether, Dibutyldiglycol

Sign Into View Organizational & Contract Pricing


About This Item

Linear Formula:
(CH3CH2CH2CH2OCH2CH2)2O
CAS Number:
Molecular Weight:
218.33
Beilstein:
1750713
EC Number:
MDL number:
UNSPSC Code:
12352100
PubChem Substance ID:
NACRES:
NA.22
form:
liquid
Assay:
≥99%
Pricing and availability is not currently available.

vapor density

>1 (vs air)

Quality Level

vapor pressure

<0.01 mmHg ( 20 °C)

Assay

≥99%

form

liquid

refractive index

n20/D 1.423 (lit.)

bp

256 °C (lit.)

mp

−60 °C (lit.)

density

0.885 g/mL at 25 °C (lit.)

functional group

ether

SMILES string

CCCCOCCOCCOCCCC

InChI

1S/C12H26O3/c1-3-5-7-13-9-11-15-12-10-14-8-6-4-2/h3-12H2,1-2H3

InChI key

KZVBBTZJMSWGTK-UHFFFAOYSA-N

Looking for similar products? Visit Product Comparison Guide

General description

The color changes in weak acid solutions of plutonium (IV) nitrate in diethylene glycol dibutyl ether has been studied[1].

Storage Class Code

10 - Combustible liquids

WGK

WGK 1

Flash Point(F)

244.4 °F - closed cup

Flash Point(C)

118 °C - closed cup

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

The properties of inorganic compounds in organic solvents?I: The hydrolysis and polymerization of quadrivalent plutonium nitrate species in diethylene glycol dibutyl ether.
Tuck DG.
J. Inorg. Nucl. Chem., 6(3), 252-256 (1958)

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service