Skip to Content
Merck
All Photos(3)

Key Documents

224111

Sigma-Aldrich

Ethylene glycol diethyl ether

98%

Synonym(s):

1,2-Diethoxyethane

Sign Into View Organizational & Contract Pricing

Select a Size

100 ML
€77.50
500 ML
€271.00

€77.50


Check Cart for Availability

Request a Bulk Order

Select a Size

Change View
100 ML
€77.50
500 ML
€271.00

About This Item

Linear Formula:
CH3CH2OCH2CH2OCH2CH3
CAS Number:
Molecular Weight:
118.17
Beilstein:
1732917
EC Number:
MDL number:
UNSPSC Code:
12352112
PubChem Substance ID:
NACRES:
NA.21
bp:
121 °C (lit.)
vapor pressure:
9.4 mmHg ( 20 °C)

€77.50


Check Cart for Availability

Request a Bulk Order

vapor pressure

9.4 mmHg ( 20 °C)

Assay

98%

form

liquid

refractive index

n20/D 1.3923 (lit.)

bp

121 °C (lit.)

mp

−74 °C (lit.)

density

0.842 g/mL at 25 °C (lit.)

SMILES string

CCOCCOCC

InChI

1S/C6H14O2/c1-3-7-5-6-8-4-2/h3-6H2,1-2H3

InChI key

LZDKZFUFMNSQCJ-UHFFFAOYSA-N

Looking for similar products? Visit Product Comparison Guide

Application


  • Correlation between urinary methoxyacetic acid and exposure of ethylene glycol dimethyl ether in a lithium battery plant.: This research investigates the exposure levels of ethylene glycol diethyl ether within a lithium battery manufacturing environment. By analyzing the correlation between urinary biomarkers and chemical exposure, the study provides insights into occupational health and safety measures required in industrial settings where such chemicals are used (Yokota et al., 2007).

Signal Word

Danger

Hazard Statements

Hazard Classifications

Eye Irrit. 2 - Flam. Liq. 2 - Repr. 1A

Supplementary Hazards

Storage Class Code

3 - Flammable liquids

WGK

WGK 2

Flash Point(F)

71.6 °F - closed cup

Flash Point(C)

22 °C - closed cup


Regulatory Listings

Regulatory Listings are mainly provided for chemical products. Only limited information can be provided here for non-chemical products. No entry means none of the components are listed. It is the user’s obligation to ensure the safe and legal use of the product.

EU REACH SVHC Candidate List

CAS No.

EU REACH Annex XVII (Restriction List)

CAS No.

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Fabrication and performance of membrane electrode assembly prepared by a catalyst-coated membrane method: effect of solvents used in a catalyst ink mixture
Therdthianwong A, et al.
Energy & Fuels, 24(2), 1191-1196 (2010)
Copper (I)-mediated living radical polymerization in the presence of oxyethylene groups: Online 1H NMR spectroscopy to investigate solvent effects
Haddleton DM, et al.
Macromolecules, 33(22), 8246-8251 (2000)
Solvate structures and computational/spectroscopic characterization of LiBF4 electrolytes
Seo DM, et al.
The Journal of Physical Chemistry C, 118(32), 18377-18386 (2014)
The bonding of fluorinated and hydrogenated diethers to Ru (001).
Walczak MM and Thiel PA.
Surface Science, 224(1), 425-450 (1989)
Fabio Marchetti et al.
Chemical communications (Cambridge, England), (31)(31), 3651-3653 (2008-07-31)
The formation of 1,4-dioxanes and alkyl chlorides from the reactions of polyethers (dme, diglyme, 1,2-diethoxyethane, 1,2-dimethoxypropane) with NbCl(5) at room temperature is described; as far as dme is concerned, the reaction mainly occurs in two steps, consisting of (i) cleavage

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service