Skip to Content
Merck
All Photos(1)

Key Documents

537527

Sigma-Aldrich

Diethylene glycol monoethyl ether acetate

99%

Synonym(s):

DE Acetate, 2-(2-Ethoxyethoxy)ethyl acetate, Ethyldiglycol acetate

Sign Into View Organizational & Contract Pricing

Select a Size

1 L
€78.60
4 L
€237.00

€78.60


Check Cart for Availability

Request a Bulk Order

Select a Size

Change View
1 L
€78.60
4 L
€237.00

About This Item

Linear Formula:
C2H5OCH2CH2OCH2CH2CO2CH3
CAS Number:
Molecular Weight:
176.21
Beilstein:
1764643
EC Number:
MDL number:
UNSPSC Code:
12352100
PubChem Substance ID:
NACRES:
NA.22

€78.60


Check Cart for Availability

Request a Bulk Order

product line

ReagentPlus®

Assay

99%

expl. lim.

8 % (lit.)

refractive index

n20/D 1.421 (lit.)

bp

218-219 °C (lit.)

mp

-25 °C (lit.)

density

1.011 g/mL at 25 °C (lit.)

functional group

ester
ether

SMILES string

CCOCCOCCOC(C)=O

InChI

1S/C8H16O4/c1-3-10-4-5-11-6-7-12-8(2)9/h3-7H2,1-2H3

InChI key

FPZWZCWUIYYYBU-UHFFFAOYSA-N

Looking for similar products? Visit Product Comparison Guide

General description

Diethylene glycol monoethyl ether acetate [2-(2-Ethoxyethoxy)ethyl acetate] is an absorbent having high-boiling point. It helps in improving the CO2 solubility as it is rich in carbonyl and ether groups.[1]

Application


  • Density and Speed of Sound of Diethylene Glycol Monoethyl Ether+ Propylene Glycol at T=(288.15 - 318.15) K: This study provides crucial thermophysical data essential for designing engineering systems involving diethylene glycol monoethyl ether (carbitol) (M Barzegar Jalali, A Jouyban, H Shekari, 2020).

  • Hydrate Equilibrium Conditions for Water, Diethylene Glycol Monoethyl Ether Acetate, and Methane: Discusses the effects of diethylene glycol monoethyl ether acetate on hydrate formation, crucial for applications in energy and environmental engineering (M Kida, M Watanabe, Y Jin, Y Konno, 2016).

Legal Information

ReagentPlus is a registered trademark of Merck KGaA, Darmstadt, Germany

Pictograms

Exclamation mark

Signal Word

Warning

Hazard Statements

Hazard Classifications

Eye Irrit. 2

Storage Class Code

10 - Combustible liquids

WGK

WGK 1

Flash Point(F)

208.4 °F - closed cup

Flash Point(C)

98 °C - closed cup

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Solubilities of CO2 capture absorbents 2-ethoxyethyl ether, 2-butoxyethyl acetate and 2-(2-ethoxyethoxy) ethyl acetate.
Li Y, et al.
Fluid Phase Equilibria, 370, 1-7 (2014)

Questions

1–2 of 2 Questions  
  1. How can I determine the shelf life / expiration / retest date of this product?

    1 answer
    1. If this product has an expiration or retest date, it will be shown on the Certificate of Analysis (COA, CofA). If there is no retest or expiration date listed on the product's COA, we do not have suitable stability data to determine a shelf life. For these products, the only date on the COA will be the release date; a retest, expiration, or use-by-date will not be displayed.
      For all products, we recommend handling per defined conditions as printed in our product literature and website product descriptions. We recommend that products should be routinely inspected by customers to ensure they perform as expected.
      For products without retest or expiration dates, our standard warranty of 1 year from the date of shipment is applicable.
      For more information, please refer to the Product Dating Information document: https://www.sigmaaldrich.com/deepweb/assets/sigmaaldrich/marketing/global/documents/449/386/product-dating-information-mk.pdf

      Helpful?

  2. How is shipping temperature determined? And how is it related to the product storage temperature?

    1 answer
    1. Products may be shipped at a different temperature than the recommended long-term storage temperature. If the product quality is sensitive to short-term exposure to conditions other than the recommended long-term storage, it will be shipped on wet or dry-ice. If the product quality is NOT affected by short-term exposure to conditions other than the recommended long-term storage, it will be shipped at ambient temperature. As shipping routes are configured for minimum transit times, shipping at ambient temperature helps control shipping costs for our customers. For more information, please refer to the Storage and Transport Conditions document: https://www.sigmaaldrich.com/deepweb/assets/sigmaaldrich/marketing/global/documents/316/622/storage-transport-conditions-mk.pdf

      Helpful?

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service