Skip to Content
Merck
All Photos(1)

Key Documents

8.43825

Sigma-Aldrich

Dipropylene glycol dimethyl ether

(mixture of isomeres)for synthesis

Synonym(s):

Dipropylene glycol dimethyl ether

Sign Into View Organizational & Contract Pricing

Select a Size

1 L
€83.50

€83.50


Check Cart for Availability

Request a Bulk Order

Select a Size

Change View
1 L
€83.50

About This Item

Empirical Formula (Hill Notation):
C8H18O3
CAS Number:
Molecular Weight:
162.23
MDL number:
UNSPSC Code:
12352112
EC Index Number:
404-640-5
NACRES:
NA.22

€83.50


Check Cart for Availability

Request a Bulk Order

vapor pressure

0.8 hPa ( 20 °C)

Quality Level

Assay

≥99.0% (GC)

form

liquid

autoignition temp.

165 °C (DIN 51794)

potency

3300 mg/kg LD50, oral (Rat)
>2000 mg/kg LD50, skin (Rat)

expl. lim.

0.77-5.57 % (v/v)

kinematic viscosity

1.12 cSt(20 °C)

bp

175 °C/1013 hPa

mp

-80 °C

transition temp

flash point 65 °C

density

0.904 g/cm3

storage temp.

2-30°C

SMILES string

O(CCCOC)CCCOC

InChI

1S/C8H18O3/c1-9-5-3-7-11-8-4-6-10-2/h3-8H2,1-2H3

InChI key

KWDCKLXGUZOEGM-UHFFFAOYSA-N

Analysis Note

Assay (GC, area%): ≥ 99.0 % (a/a)
Density (d 20 °C/ 4 °C): 0.902 - 0.906
Identity (IR): passes test

Storage Class Code

10 - Combustible liquids

WGK

WGK 1

Flash Point(F)

149.0 °F - closed cup

Flash Point(C)

65 °C - closed cup


Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service