Skip to Content
Merck
All Photos(2)

Key Documents

11955

Sigma-Aldrich

Bengal Rose B bound to Polystyrene

200-400 mesh, ~0.1 mmol/g, resin loading

Synonym(s):

Rose bengal B bound to polystyrene

Sign Into View Organizational & Contract Pricing

Select a Size

1 G
€285.00

€285.00


Estimated to ship on07 April 2025


Request a Bulk Order

Select a Size

Change View
1 G
€285.00

About This Item

MDL number:
UNSPSC Code:
23151817
PubChem Substance ID:
NACRES:
NA.22

€285.00


Estimated to ship on07 April 2025


Request a Bulk Order

reaction suitability

reaction type: solution phase peptide synthesis

matrix

crosslinked with 1% DVB

particle size

200-400 mesh

capacity

~0.1 mmol/g, resin loading

Other Notes

Heterogenous photo-sensitizer for the generation of singlet oxygen used in photooxygenation reactions[1][2][3][4]
Sales restrictions may apply

Storage Class Code

11 - Combustible Solids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

A.P. Schaap et al.
Tetrahedron Letters, 28, 1159-1159 (1987)
J.J. Lamberts et al.
Journal of the American Chemical Society, 105, 7465-7465 (1983)
N.V. Shinkarenko et al.
Russ. Chem. Rev., 50, 220-220 (1981)
R.S. Handley et al.
Tetrahedron Letters, 26, 3183-3183 (1985)

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service