Skip to Content
Merck
All Photos(2)

Key Documents

900855

Sigma-Aldrich

1,1,2,2-Tetrakis(4-bromophenyl)ethylene

Synonym(s):

1,1′,1′′,1′′′-(1,2-Ethenediylidene)tetrakis[4-bromobenzene]

Sign Into View Organizational & Contract Pricing

Select a Size

500 MG
€240.00

€240.00


Check Cart for Availability

Request a Bulk Order

Select a Size

Change View
500 MG
€240.00

About This Item

Empirical Formula (Hill Notation):
C26H16Br4
CAS Number:
Molecular Weight:
648.02
MDL number:
UNSPSC Code:
12352103
NACRES:
NA.23

€240.00


Check Cart for Availability

Request a Bulk Order

form

powder or crystals

Quality Level

solubility

DMF: soluble
DMSO: soluble
THF: soluble
dichloromethane: soluble

storage temp.

2-8°C

SMILES string

Brc1ccc(cc1)C(=C(c4ccc(cc4)Br)c3ccc(cc3)Br)c2ccc(cc2)Br

InChI

1S/C26H16Br4/c27-21-9-1-17(2-10-21)25(18-3-11-22(28)12-4-18)26(19-5-13-23(29)14-6-19)20-7-15-24(30)16-8-20/h1-16H

InChI key

BIRLDGKMJJEZRI-UHFFFAOYSA-N

General description

AIE (aggregation-induced emission)-active building block.

Storage Class Code

11 - Combustible Solids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Nathan D Rudd et al.
ACS applied materials & interfaces, 8(44), 30294-30303 (2016-10-14)
We have designed and synthesized an isoreticular series of luminescent metal-organic frameworks (LMOFs) by incorporating a strongly emissive molecular fluorophore and functionally diverse colinkers into Zn-based structures. The three-dimensional porous networks of LMOF-261, -262, and -263 represent a unique/new type
Natalia B Shustova et al.
Journal of the American Chemical Society, 133(50), 20126-20129 (2011-11-15)
Coordinative immobilization of functionalized tetraphenylethylene within rigid porous metal-organic frameworks (MOFs) turns on fluorescence in the typically non-emissive tetraphenylethylene core. The matrix coordination-induced emission effect (MCIE) is complementary to aggregation-induced emission. Despite the large interchromophore distances imposed by coordination to
Sasanka Dalapati et al.
Journal of the American Chemical Society, 138(18), 5797-5800 (2016-04-26)
Highly luminescent covalent organic frameworks (COFs) are rarely achieved because of the aggregation-caused quenching (ACQ) of π-π stacked layers. Here, we report a general strategy to design highly emissive COFs by introducing an aggregation-induced emission (AIE) mechanism. The integration of

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service