Skip to Content
Merck
All Photos(2)

Key Documents

T26204

Sigma-Aldrich

1,1,2,2-Tetraphenylethylene

1,1,2,2-Tetraphenylethylene

98%

Synonym(s):

α,β-Diphenylstilbene, 1,1,2,2-Tetraphenylethene, 1,1′,1′′,1′′′-(1,2-Ethenediylidene)tetrakis[benzene], 1,2,2-Triphenylethenylbenzene, Tetraphenylethene

Sign Into View Organizational & Contract Pricing

Select a Size

5 G
€85.70

€85.70


Check Cart for Availability

Request a Bulk Order

Select a Size

Change View
5 G
€85.70

About This Item

Linear Formula:
(C6H5)2C=C(C6H5)2
CAS Number:
Molecular Weight:
332.44
Beilstein:
1913548
EC Number:
MDL number:
UNSPSC Code:
12352100
PubChem Substance ID:
NACRES:
NA.22

€85.70


Check Cart for Availability

Request a Bulk Order

Assay

98%

bp

420 °C (lit.)

mp

222-224 °C (lit.)

SMILES string

c1ccc(cc1)\C(c2ccccc2)=C(\c3ccccc3)c4ccccc4

InChI

1S/C26H20/c1-5-13-21(14-6-1)25(22-15-7-2-8-16-22)26(23-17-9-3-10-18-23)24-19-11-4-12-20-24/h1-20H

InChI key

JLZUZNKTTIRERF-UHFFFAOYSA-N

Looking for similar products? Visit Product Comparison Guide

Storage Class Code

11 - Combustible Solids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Cheng Wang et al.
Acta biomaterialia, 85, 218-228 (2018-12-18)
In this study, an AIE-active polymer (FTP) was successfully prepared and employed to load anti-cancer drug doxorubicin (DOX) for self-indicating cancer therapy via dual FRET process. Our results demonstrated that the FTP polymer could self-assemble into nanoparticles (NPs) in aqueous
Chang-Rui Guo et al.
ACS omega, 6(3), 2177-2183 (2021-02-02)
The introduction of nitrogen-rich functional groups into a luminescent metal-organic framework (LMOF) can enhance its fluorescent sensing ability. In this work, we designed and synthesized a triazole-containing tetracarboxyl-substituted tetraphenylethene (TPE) ligand, tetrakis[4-(4-carboxyphenyl)(1H-1,2,3-triazol-4,1-diyl)phenyl]ethene (H4TCPTAPE), featuring a prominent aggregation-induced emission (AIE). A
Raúl Villamil-Ramos et al.
Analytica chimica acta, 1057, 51-59 (2019-03-06)
Sensing of pyrophosphate anion (PPi) in the presence of nucleotide triphosphates allows the real time monitoring of the polymerase chain reaction. To get a deeper understanding of the factors involved in PPi/nucleotide triphosphate discrimination, a detailed study on the performance
Sandeep Urandur et al.
ACS applied materials & interfaces, 10(15), 12960-12974 (2018-03-27)
The prospective design of nanocarriers for personalized oncotherapy should be an ensemble of targeting, imaging, and noninvasive therapeutic capabilities. Herein, we report the development of the inverse hexagonal nano-liquid crystalline (NLC) particles that are able to host formononetin (FMN), a
Seiji Tsunega et al.
ChemPlusChem, 85(4), 619-626 (2020-04-03)
Systems that show circularly polarized luminescence (CPL) are usually constructed in one of two possible ways: either covalently binding the chiral moieties (usually organic compounds) to luminophores (inorganic or organic compounds) or associating the luminophores as guests with chiral hosts

Articles

AIE Luminogens - Family of new materials with multifaceted functionalities. Find some representative applications of the AIEgens

Questions

1–2 of 2 Questions  
  1. How can I determine the shelf life / expiration / retest date of this product?

    1 answer
    1. If this product has an expiration or retest date, it will be shown on the Certificate of Analysis (COA, CofA). If there is no retest or expiration date listed on the product's COA, we do not have suitable stability data to determine a shelf life. For these products, the only date on the COA will be the release date; a retest, expiration, or use-by-date will not be displayed.
      For all products, we recommend handling per defined conditions as printed in our product literature and website product descriptions. We recommend that products should be routinely inspected by customers to ensure they perform as expected.
      For products without retest or expiration dates, our standard warranty of 1 year from the date of shipment is applicable.
      For more information, please refer to the Product Dating Information document: https://www.sigmaaldrich.com/deepweb/assets/sigmaaldrich/marketing/global/documents/449/386/product-dating-information-mk.pdf

      Helpful?

  2. How is shipping temperature determined? And how is it related to the product storage temperature?

    1 answer
    1. Products may be shipped at a different temperature than the recommended long-term storage temperature. If the product quality is sensitive to short-term exposure to conditions other than the recommended long-term storage, it will be shipped on wet or dry-ice. If the product quality is NOT affected by short-term exposure to conditions other than the recommended long-term storage, it will be shipped at ambient temperature. As shipping routes are configured for minimum transit times, shipping at ambient temperature helps control shipping costs for our customers. For more information, please refer to the Storage and Transport Conditions document: https://www.sigmaaldrich.com/deepweb/assets/sigmaaldrich/marketing/global/documents/316/622/storage-transport-conditions-mk.pdf

      Helpful?

Reviews

Active Filters

  1. Glasgow, UK
    • Review 1
    • Vote 1
    5 out of 5 stars.

    very satisfactory

    It met the expectations. The quality was good and high standard.

    Helpful?

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service