Skip to Content
Merck
All Photos(1)

Key Documents

R1273

Millipore

Rose bengal agar base

suitable for microbiology, NutriSelect® Plus

Sign Into View Organizational & Contract Pricing

Select a Size

500 G
€223.00

€223.00


Check Cart for Availability

Request a Bulk Order

Select a Size

Change View
500 G
€223.00

About This Item

UNSPSC Code:
41171606
NACRES:
NA.85

€223.00


Check Cart for Availability

Request a Bulk Order

sterility

non-sterile

Quality Level

form

powder

shelf life

limited shelf life, expiry date on the label

manufacturer/tradename

NutriSelect® Plus

technique(s)

microbiological culture: suitable

pH

7.2±0.2 (25 °C)

application(s)

bioburden testing
clinical testing
environmental
food and beverages

microbiology

suitability

selective for Aspergillus spp.
selective for Candida spp.
selective for Penicillium spp.
selective for Pichia spp.
selective for Saccharomyces spp.
selective for Zygosaccharomyces spp.
selective for molds (General Media)
selective for yeasts (General Media)

Application

Ingredients (g/L) for the selective isolation and enumeration of yeasts and molds.

Components

Ingredients (g/L)
Papic digest of soybean meal, 5.00
Dextrose, 10.00
Monopotassium phosphate, 1.00
Magnesium sulphate, 0.50
Rose bengal, 0.05
Agar, 15.00

Preparation Note

Suspend 31.55 g of Rose Bengal Agar Base in 1000 ml of distilled water. Boil to dissolve the medium completely. Sterilize by autoclaving at 15 lbs. pressure (121°C) for 15 minutes. Cool to 45°C and add 2 vials of rehydrated Chloramphenicol Selective Supplement (Cat. No. 29231) to 1 litre of the Rose Bengal Agar Base. Mix thoroughly and pour into sterile petri plates.

Footnote

We offer two media types: the superior granulated GranuCult® and the cost-efficient powdered NutriSelect® culture media, depending on your needs.
The designations basic, plus, or prime are added to indicate the quality control level, from basic quality control to standard QC plus to prime for full regulatory compliance.

Legal Information

GRANUCULT is a registered trademark of Merck KGaA, Darmstadt, Germany
NutriSelect is a registered trademark of Merck KGaA, Darmstadt, Germany

Storage Class Code

11 - Combustible Solids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Comparison of an improved rose bengal-chlortetracycline agar with other media for the selective isolation and enumeration of moulds and yeasts in foods.
B Jarvis
The Journal of applied bacteriology, 36(4), 723-727 (1973-12-01)

Articles

Yeast is one of the most important microorganisms known and utilised by mankind. Ancient Middle Eastern civilisations used the organism to bake bread and to produce mead, beer and wine.

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service