Merck
Millipak® Express 20 Filter 0.22 µm membrane filter for particulate-free and bacteria-free water at the point of dispense, For use with Direct-Q®, Synergy® and Milli-Q® Academic / Biocel / Element / Synthesis / Gradient systems
Millipak® Express 40 Filter
Millipak® Express 20 Filter 0.22 µm membrane filter for particulate-free and bacteria-free water at the point of dispense, For use with Direct-Q®, Synergy® and Milli-Q® Academic / Biocel / Element / Synthesis / Gradient systems
Synonym(e):
Millipak® Express Filter
Products
Produktkategorie
Marke
Kompatibilität
Qualitätsstufe
Gehäusematerial
Porengröße (µm)
Sterilität