Größe auswählen
| Größe/SKU | Verfügbarkeit | Preis |
|---|
Über diesen Artikel
material
Delrin® nut, Tefzel® ferrule (flangeless)
packaging
pkg of 5 ea
parameter
500 psi max. pressure (35 bar)
fitting
1/4-28 male Upchurch nut and ferrule for 1/8 in. O.D. tubing
General description
Legal Information
1 of 1
Dieser Artikel | |||
|---|---|---|---|
| description 1/4-28 male Upchurch nut and ferrule for 1/8 in. O.D. tubing | description 1/4-28 male Upchurch nut and ferrule for 1/16 in. O.D. tubing | description for tubing i.d., 1/4 in. | description 1/4-28 male UNF for 1/8 in. O.D. tubing 3 mm (includes washer), (natural color) |
| fitting 1/4-28 male Upchurch nut and ferrule for 1/8 in. O.D. tubing | fitting 1/4-28 male Upchurch nut and ferrule for 1/16 in. O.D. tubing | fitting - | fitting 1/4-28 male UNF for 1/8 in. O.D. tubing 3 mm (includes washer) |
| material Delrin® nut, Tefzel® ferrule (flangeless) | material Delrin® nut, Tefzel® ferrule (flangeless) | material - | material (natural color), stainless steel washer, natural polypropylene nut |
| parameter 500 psi max. pressure (35 bar) | parameter 1000 psi max. pressure (70 bar) | parameter - | parameter - |
| packaging pkg of 5 ea | packaging pkg of 5 ea | packaging - | packaging pkg of 10 ea |
Hier finden Sie alle aktuellen Versionen:
Besitzen Sie dieses Produkt bereits?