33281
Eigenschaften
Decapper, pliers style
pkg of 1 ea
for use with 11 mm crimp seals
Beschreibung
Unterschiede anzeigen
1 of 1
Fläschchenöffner, Zange
33282
33283
Sigma-Aldrich
Z114278
Decapper-Zange
for use with 11 mm crimp seals, pkg of 1 ea
for use with 13mm crimp seals, pkg of 1 ea
for use with 20mm crimp seals, pkg of 1 ea
O.D. 20 mm
for use with 13mm crimp seals
for use with 20mm crimp seals
-
Explore all of our products under Fläschchenöffner, Zange
Hier finden Sie alle aktuellen Versionen:
It looks like we've run into a problem, but you can still download Certificates of Analysis from our Dokumente section.