Größe auswählen
| Größe/SKU | Verfügbarkeit | Preis |
|---|
Über diesen Artikel
material
stainless steel
packaging
pkg of 2 ea
manufacturer/tradename
SWAGELOK® 201-PC
technique(s)
gas chromatography (GC): suitable using :Suitable
fitting
1/8 in. SWAGELOK
Application
Legal Information
1 of 1
Dieser Artikel | |||
|---|---|---|---|
| description SWAGELOK® 201-PC, stainless steel, 1/8 in. SWAGELOK | description SWAGELOK® 201-PC, brass, 1/8 in. SWAGELOK | description brass, 1/8 in. SWAGELOK, 1/8 in. male NPT, 200-1-2 | description brass, 1/4 in. SWAGELOK, 1/8 in. male NPT, 400-1-2 |
| material stainless steel | material brass | material brass | material brass |
| manufacturer/tradename SWAGELOK® 201-PC | manufacturer/tradename SWAGELOK® 201-PC | manufacturer/tradename 200-1-2 | manufacturer/tradename 400-1-2 |
| packaging pkg of 2 ea | packaging pkg of 2 ea | packaging pkg of 2 ea | packaging pkg of 2 ea |
| technique(s) gas chromatography (GC): suitable using :Suitable | technique(s) - | technique(s) gas chromatography (GC): suitable using :Suitable | technique(s) gas chromatography (GC): suitable using :Suitable |
| fitting 1/8 in. SWAGELOK | fitting 1/8 in. SWAGELOK | fitting 1/8 in. SWAGELOK, 1/8 in. male NPT | fitting 1/8 in. male NPT, 1/4 in. SWAGELOK |
Still not finding the right product?
Explore all of our products under Swagelok® Anschlussstecker
Hier finden Sie alle aktuellen Versionen:
Besitzen Sie dieses Produkt bereits?