Größe auswählen
| Größe/SKU | Verfügbarkeit | Preis |
|---|---|---|
1 ea | Warenkorb auf Verfügbarkeit prüfen | € 52,20 |
Über diesen Artikel
€ 52,20
description
15 mL QuEChERS Extraction Tube Shaker Rack
feature
Holds 50 x 15 mL tubes
packaging
pkg of 1 ea
manufacturer/tradename
Benchmark Scientific BV1010-150
General description
Legal Information
1 of 1
Dieser Artikel | |||
|---|---|---|---|
| description Holds 50 x 15 mL tubes | description Holds 15 x 25 mL tubes | description 3 x 96 x 0.2 mL tube/strip carrier | description Holds 5 x 50 mL tubes |
| manufacturer/tradename Benchmark Scientific BV1010-150 | manufacturer/tradename Benchmark Scientific BV1010-25 | manufacturer/tradename Benchmark Scientific BV1010-MP02 | manufacturer/tradename Benchmark Scientific BV1003-T500 |
| feature Holds 50 x 15 mL tubes | feature Holds 15 x 25 mL tubes | feature 3 x 96 x 0.2 mL tube/strip carrier | feature Holds 5 x 50 mL tubes |
| packaging pkg of 1 ea | packaging pkg of 1 ea | packaging pkg of 1 ea | packaging pack of 1 ea |
| description 15 mL QuEChERS Extraction Tube Shaker Rack | description - | description - | description - |
Still not finding the right product?
Explore all of our products under Tube rack for Benchmixer™ XL multi-tube vortexer
Hier finden Sie alle aktuellen Versionen:
Besitzen Sie dieses Produkt bereits?