Fortfahren mit
Z764094
MicroTissues® 3D Petri Dish® micro-mold mixed spheroid kit
3 small and 3 large, fits 12 well plates
Aktionsangebot verfügbar
Synonym(e):
3D, 3D Cell Culture, Microtissues 12-1
Anmelden zur Ansicht der Organisations- und Vertragspreise.
Spend € 450 — Save 5 %; Spend € 700 — Save 10 %; Spend € 1.500 — Save 15 %*
Anmelden zur Überprüfung der Spielberechtigung. Produkte kaufen
Größe auswählen
Ansicht ändern
Größe/SKUVerfügbarkeitPreis
6 ea
Warenkorb auf Verfügbarkeit prüfen
€ 638,00
Über diesen Artikel
NACRES:
NB.13
UNSPSC Code:
41121812
Material:
spherical
Sterility:
sterile; autoclaved
Manufacturer/tradename:
MicroTissues Inc. 12-1
Packaging:
pack of (Contains 3 small and 3 large)
€ 638,00
Sparen Sie bis zu 15 %
Warenkorb auf Verfügbarkeit prüfen
Technischer Dienst
Benötigen Sie Hilfe? Unser Team von erfahrenen Wissenschaftlern ist für Sie da.
Unterstützung erhalten
material
spherical
sterility
sterile; autoclaved
feature
lid: no
manufacturer/tradename
MicroTissues Inc. 12-1
packaging
pack of (Contains 3 small and 3 large)
technique(s)
cell culture: suitable
application(s)
3D cell culture
General description
Mixed spheroid pack for 12-well plates. Mixed pack of six autoclavable precision micro-molds to cast 3D Petri Dish for forming small or large spheroids. 3D Petri Dish for use in 12-well plate. Contains 3 x Z764000 micro-molds for making small spheroids and 3 x Z764019 micro-molds for making larger spheroids. Perfect for determining which size spheroid is right for your experiments.
When the gelled agarose is removed from the micro-mold, it is transferred to a standard 12 well or 24 well tissue culture dish and equilibrated with cell culture medium.
Since the agarose is transparent, the spheroids or microtissues that form at the bottom of each agarose micro-well can be easily viewed using a standard inverted microscope using phase contrast, bright field or fluorescence microscopy.
Features and Benefits
  • The micro-molds are reusable up to 12 times and can be sterilized via a standard steam autoclave (30 min, dry cycle).
  • The micro-molds should be stored in a covered container to maintain sterility and to avoid collecting dust or fibres on their small features.
  • Any microscope with sufficient magnification and resolution functions for cell based views or images will be suitable for use with the 3D Petri Dish products.
Legal Information
3D Petri Dish is a registered trademark of MicroTissues Inc.
MicroTissues is a registered trademark of MicroTissues Inc.
Ähnliche Artikel vergleichen
Vollständigen Vergleich anzeigen
Unterschiede anzeigen
1 of 1
Dieser Artikel
Z764043Z764019Z764078
description
3 small and 3 large, fits 12 well plates
description
size S, 8 x 12 array, fits 24 well plates
description
size L, 9 x 9 array, fits 12 well plates
description
fits 24 well plates, 3 x 8 array
sterility
sterile; autoclaved
sterility
sterile; autoclaved
sterility
sterile; autoclaved
sterility
sterile; autoclaved
manufacturer/tradename
MicroTissues Inc. 12-1
manufacturer/tradename
MicroTissues Inc. 24-96
manufacturer/tradename
MicroTissues Inc. 12-81
manufacturer/tradename
MicroTissues Inc. 24-24TR
feature
lid: no
feature
8 x 12 array, lid: no
feature
lid: no, 9 x 9 array
feature
3 x 8 array, lid: no
material
spherical
material
spherical
material
spherical
material
spherical
packaging
pack of (Contains 3 small and 3 large)
packaging
pack of 6 ea
packaging
pack of 6 ea
packaging
pack of 6 ea
Still not finding the right product?
Hier finden Sie alle aktuellen Versionen:
Analysenzertifikate (COA)