Größe auswählen
| Größe/SKU | Verfügbarkeit | Preis |
|---|---|---|
6 ea | Warenkorb auf Verfügbarkeit prüfen | € 638,00 |
Über diesen Artikel
€ 638,00
material
spherical
description
makes rod-shaped microtissues
sterility
sterile; autoclaved
feature
lid: no, 3 x 8 array
packaging
pack of 6 ea
manufacturer/tradename
MicroTissues Inc. 24-24TR
technique(s)
cell culture: suitable
volume
200 μL
application(s)
3D cell culture
General description
- Nominal dimensions of each 3D culture trough: L 2200 μm x D 800 μm x W 400 μm
- Micro-molds also form single chamber for cell seeding
Since the agarose is transparent, the spheroids or microtissues that form at the bottom of each agarose micro-well can be easily viewed using a standard inverted microscope using phase contrast, bright field or fluorescence microscopy.
- The micro-molds are reusable up to 12 times and can be sterilized via a standard steam autoclave (30 min, dry cycle)
- The micro-molds should be stored in a covered container to maintain sterility and to avoid collecting dust or fibres on their small features
- Any microscope with sufficient magnification and resolution functions for cell based views or images will be suitable for use with the 3D Petri Dish products
Legal Information
1 of 1
Dieser Artikel | |||
|---|---|---|---|
| description fits 24 well plates, 3 x 8 array | description fits 12 well plates, 4 x 15 array | description size S, 8 x 12 array, fits 24 well plates | description makes toroid-shaped microtissues, fits 12 well plates, 6 x 6 array |
| sterility sterile; autoclaved | sterility sterile; autoclaved | sterility sterile; autoclaved | sterility sterile; autoclaved |
| feature 3 x 8 array, lid: no | feature 4 x 15 array, lid: no | feature 8 x 12 array, lid: no | feature 6 x 6 array, lid: no |
| manufacturer/tradename MicroTissues Inc. 24-24TR | manufacturer/tradename MicroTissues Inc. 12-60TR | manufacturer/tradename MicroTissues Inc. 24-96 | manufacturer/tradename MicroTissues Inc. 12-36TO |
| material spherical | material spherical | material spherical | material spherical |
| packaging pack of 6 ea | packaging pack of 6 ea | packaging pack of 6 ea | packaging pack of 6 ea |
| description makes rod-shaped microtissues | description makes rod-shaped microtissues | description - | description - |
Still not finding the right product?
Explore all of our products under MicroTissues® 3D Petri Dish® micro-mold rods
Hier finden Sie alle aktuellen Versionen:
Besitzen Sie dieses Produkt bereits?