Fortfahren mit
€ 450 ausgeben – 5 % sparen; € 700 ausgeben – 10 % sparen; € 1.500 ausgeben – 15 % sparen*
Anmelden zur Überprüfung der Spielberechtigung. Produkte kaufen
Größe auswählen
| Größe/SKU | Verfügbarkeit | Preis |
|---|---|---|
960 ea | Warenkorb auf Verfügbarkeit prüfen | € 193,00 |
Über diesen Artikel
€ 193,00
Sparen Sie bis zu 15 %
material
colorless , polypropylene (Virgin)
sterility
sterile
feature
DNase free, RNase free, barrier, filter
packaging
pack of 960 ea (10 racks of 96 tips)
manufacturer/tradename
Sartorius 790201F
maximum volume
200 μL
volume range
5-200 μL
volume
5—200 μL
binding type
low binding surface
General description
These tips have more space between the sample and the filter than conventional filter tips so you do not need to worry about the sample touching the filter whatever the pipetting technique or type of liquid.
Benefits:
- Pipette foaming liquids like buffers and proteins
- Pipette solvents
- Reverse pipetting
- Multiple dispensing functions of electronic pipettes
Other Notes
1 of 1
Dieser Artikel | |||
|---|---|---|---|
| description volume range 5-200 μL, sterile, pack of 960 ea (10 racks of 96 tips) | description volume range 0.5-20 μL, sterile, pack of 960 ea (10 racks of 96 tips) | description volume range 0.1-10 μL, sterile, pack of 960 ea (10 racks of 96 tips) | description volume range 50-1000 μL, sterile, pack of 960 ea (10 racks of 96 tips) |
| feature DNase free, filter, barrier, RNase free | feature DNase free, filter, barrier, RNase free | feature barrier, RNase free, DNase free, filter | feature barrier, RNase free, DNase free, filter |
| volume 5—200 μL | volume 0.5—20 μL | volume 0.1—10 μL | volume 50—1000 μL |
| manufacturer/tradename Sartorius 790201F | manufacturer/tradename Sartorius 790021F | manufacturer/tradename Sartorius 790011F | manufacturer/tradename Sartorius 791001F |
| sterility sterile | sterility sterile | sterility sterile | sterility sterile |
| material colorless , polypropylene (Virgin) | material polypropylene (Virgin), colorless | material polypropylene (Virgin), colorless | material polypropylene (Virgin), colorless , colorless tip |
| binding type low binding surface | binding type low binding surface | binding type low binding surface | binding type low binding surface |
Still not finding the right product?
Explore all of our products under Sartorius filter pipette tips
Hier finden Sie alle aktuellen Versionen:
Besitzen Sie dieses Produkt bereits?