Fortfahren mit
Z757683
Sartorius pipette tips
volume range 0.5-200 μL, standard, racked, sterile
Anmelden zur Ansicht der Organisations- und Vertragspreise.
Größe auswählen
Ansicht ändern
Größe/SKUVerfügbarkeitPreis
960 ea
Warenkorb auf Verfügbarkeit prüfen
€ 101,00
Über diesen Artikel
NACRES:
NB.01
UNSPSC Code:
41121600
eCl@ss:
32039090
Volume:
0.5—200 μL
Sterility:
sterile
Feature:
DNase free, RNase free, barrier: no, endotoxin free
Packaging:
pack of 96 ea (Racked)
Manufacturer/tradename:
Sartorius 790201
€ 101,00
Warenkorb auf Verfügbarkeit prüfen
​
Technischer Dienst
Benötigen Sie Hilfe? Unser Team von erfahrenen Wissenschaftlern ist für Sie da.
Unterstützung erhalten
material
colorless
sterility
sterile
feature
DNase free, RNase free, barrier: no, endotoxin free
packaging
pack of 96 ea (Racked)
manufacturer/tradename
Sartorius 790201
maximum volume
200 μL
volume range
0.5-200 μL
volume
0.5—200 μL
General description
The non-filtered Optifit Tips are designed for best possible fitting and sealing to secure the highest possible accuracy and precision. These pipette tips enable ergonomic and light tip attachment.
Optifit Tips are available in various packaging types for all your needs:
  • Single Tray racks – purity certified tip racks
  • Refill Packs - purity certified and ecological refill packages
  • Refill Towers - space-saving and ecological refill packages
  • Flexibulk boxes - space-saving, ecological and economical bulk boxes
  • Standard bulk – economical bulk boxes
While Optifit Tip trays are colour coded for easy matching with a corresponding, colour coded Sartorius pipette, they do also match with most other pipette due to their universal design.
Strict quality standards, ISO 9001 and ISO 13485, are followed from tip R&D to production and delivery. The purity certified tips are manufactured in ISO 8 classified clean room conditions to avoid any contaminations and are tested for DNase, RNase and endotoxins per each tip lot. Pre-sterilised tips are also available for the purest laboratory work. Sartorius offers a selection of pipette tips, extended and large volume tips, for special needs.
Other Notes
Browse all pipette tips
Ähnliche Artikel vergleichen
Vollständigen Vergleich anzeigen
Unterschiede anzeigen
1 of 1
Dieser Artikel
Z757705Z709867Z709972
description
volume range 0.5-200 μL, standard, racked, sterile
description
volume range 0.5-200 μL, standard, refill, non-sterile
description
volume range 0.2-10 μL, standard, racked, sterile
description
volume range 10-1000 μL, standard, refill, non-sterile
manufacturer/tradename
Sartorius 790201
manufacturer/tradename
Sartorius 790202
manufacturer/tradename
Sartorius 790011
manufacturer/tradename
Sartorius 791002
feature
DNase free, endotoxin free, RNase free, barrier: no
feature
barrier: no, endotoxin free, DNase free, RNase free
feature
RNase free, endotoxin free, barrier: no, DNase free
feature
RNase free, DNase free, endotoxin free, barrier: no
volume
0.5—200 μL
volume
0.5—200 μL
volume
10 μL
volume
10—1000 μL
material
colorless
material
colorless
material
colorless
material
colorless
packaging
pack of 96 ea (Racked)
packaging
pack of 96 ea (Racked)
packaging
pack of 96 ea (racked)
packaging
pack of 96 ea (racked)
volume range
0.5-200 μL
volume range
0.5-200 μL
volume range
-
volume range
-
Still not finding the right product?
Explore all of our products under Sartorius pipette tips
Hier finden Sie alle aktuellen Versionen:
Analysenzertifikate (COA)