Ansicht ändern
| Ihnen/SKU | Verfügbarkeit | Preis |
|---|---|---|
500 ea | Warenkorb auf Verfügbarkeit prüfen | € 36,50 € 32,85 |
Über diesen Artikel
NACRES:
NB.35
UNSPSC Code:
41122410
Manufacturer/tradename:
Diversified Biotech FOIL-2000
Sterility:
non-sterile
Packaging:
pack of 500 sheets
€ 32,85
Listenpreis€ 36,50Sparen Sie 10%Aktion nur im Web
Warenkorb auf Verfügbarkeit prüfen
Technischer Dienst
Benötigen Sie Hilfe? Unser Team von erfahrenen Wissenschaftlern ist für Sie da.
Unterstützung erhaltensterility
non-sterile
packaging
pack of 500 sheets
manufacturer/tradename
Diversified Biotech FOIL-2000
L × W
3 in. × 3 in.
General description
Speedy-Foil is pre-cut aluminum squares interleved with tissue paper for conveniently and quickly covering lab vessels.
Features and Benefits
- Faster, easier and aseptic: grab and cover
- Pre-cut to fit common laboratory containers
- No scissors and less waste
- 1 mil thick for a sharp, crisp dead-fold providing a tighter fit
- Each square is interleaved for physical separation
- Meets FDA requirements for direct contact with pharmaceuticals
- RNase and DNase free
Legal Information
Speedy-Foil is a registered trademark of Diversified Biotech
1 of 1
Dieser Artikel | |||
|---|---|---|---|
| description L × W 3 in. × 3 in., pack of 500 sheets | description L × W 4 in. × 4 in., pack of 500 sheets | description L × W 5 in. × 5 in., pack of 500 sheets | description W 4 in. (102 mm) |
| sterility non-sterile | sterility non-sterile | sterility non-sterile | sterility non-sterile |
| manufacturer/tradename Diversified Biotech FOIL-2000 | manufacturer/tradename Diversified Biotech FOIL-3000 | manufacturer/tradename Diversified Biotech FOIL-4000 | manufacturer/tradename Heathrow Scientific HS23534A |
| packaging pack of 500 sheets | packaging pack of 500 sheets | packaging pack of 500 sheets | packaging - |
| L × W 3 in. × 3 in. | L × W 4 in. × 4 in. | L × W 5 in. × 5 in. | L × W - |
Still not finding the right product?
Explore all of our products under Speedy-Foil® aluminium squares interleaved with paper
Hier finden Sie alle aktuellen Versionen:
Besitzen Sie dieses Produkt bereits?