Ansicht ändern
| Ihnen/SKU | Verfügbarkeit | Preis |
|---|---|---|
2 ea | Warenkorb auf Verfügbarkeit prüfen | € 42,70 |
Über diesen Artikel
NACRES:
NB.35
UNSPSC Code:
55121611
€ 42,70
Warenkorb auf Verfügbarkeit prüfen
Technischer Dienst
Benötigen Sie Hilfe? Unser Team von erfahrenen Wissenschaftlern ist für Sie da.
Unterstützung erhaltenmanufacturer/tradename
Diversified Biotech VIBE-3000
packaging
pkg of One of each size
L × W
7.0 in. × 8.5 in. , small, 8.5 in. × 11.0 in. , large
General description
Inexpensive solution which dampens vibrations generated from lab equipment
Creates a stable surface to keep equipment in place
Fits vortexes, shakers, rockers and microscopes
Small, lightweight design
Stack pads for heavier units or position side-by-side forlarger equipment
Creates a stable surface to keep equipment in place
Fits vortexes, shakers, rockers and microscopes
Small, lightweight design
Stack pads for heavier units or position side-by-side forlarger equipment
1 of 1
Dieser Artikel | |||
|---|---|---|---|
| description combi pack, L × W 7.0 in. × 8.5 in. , small, L × W 8.5 in. × 11.0 in. , large, pkg of One of each size | description L × W 7.0 in. × 8.5 in., pkg of 2 | description L × W 8.5 in. × 11.0 in., pkg of 2 | description size 18 in. × 22 in. |
| packaging pkg of One of each size | packaging pkg of 2 | packaging pkg of 2 | packaging pack of 1 ea |
| manufacturer/tradename Diversified Biotech VIBE-3000 | manufacturer/tradename Diversified Biotech VIBE-1000 | manufacturer/tradename Diversified Biotech VIBE-2000 | manufacturer/tradename Bel-Art H18386-0000 |
| L × W 7.0 in. × 8.5 in. , small | L × W 7.0 in. × 8.5 in. | L × W 8.5 in. × 11.0 in. | L × W - |
Still not finding the right product?
Explore all of our products under Richter Anti-Vibration pads
Hier finden Sie alle aktuellen Versionen:
Besitzen Sie dieses Produkt bereits?