Fortfahren mit
Z742383
CellCrown™ inserts
12 well plate inserts, sterile
Synonym(e):
3D cell inserts, Scaffdex C00007S
Anmelden zur Ansicht der Organisations- und Vertragspreise.
Größe auswählen
Ansicht ändern
Größe/SKUVerfügbarkeitPreis
6 ea
Warenkorb auf Verfügbarkeit prüfen
€ 63,00
Über diesen Artikel
NACRES:
NB.22
UNSPSC Code:
41121800
eCl@ss:
32040503
Material:
clear polycarbonate , polycarbonate
Sterility:
sterile; γ-irradiated, sterile
€ 63,00
Warenkorb auf Verfügbarkeit prüfen
​
Technischer Dienst
Benötigen Sie Hilfe? Unser Team von erfahrenen Wissenschaftlern ist für Sie da.
Unterstützung erhalten
material
clear polycarbonate , polycarbonate
sterility
sterile; γ-irradiated, sterile
feature
lid
packaging
pack of 6 ea
manufacturer/tradename
Scaffdex C00007S
size
12 well
application(s)
3D cell culture
General description
CellCrown is a disposable insert, which immobilizes a sample in its place on a 6, 12, 24, 48, and 96-well plate. Fibers, foils, membranes, textiles, tissue and block like samples can be fixed with CellCrown in a firm, adjusted position in the well. The sample can be left in the well during the change of media by means of a pipette between the wall of the well and the CellCrown. The lid of the well plate can be closed while CellCrown is used. The insert can be removed from the well plate and unloaded after cell culturing without damaging the sample. CellCrown is made of medical grade polycarbonate and the product range includes sterile products ready to use as well as non-sterile products to be sterilized by the user. Sterilization can be by gamma or e-beam radiation, autoclaving, ethylene oxide gas and plasma sterilization.
Sterilization Conditions:
Temperature.......121 °C / 250 °F
Time..................15 min.
Drying time.........15 min.
Legal Information
CellCrown is a trademark of Scaffdex Oy
Ähnliche Artikel vergleichen
Vollständigen Vergleich anzeigen
Unterschiede anzeigen
1 of 1
Dieser Artikel
Z742380Z681822Z682004
description
12 well plate inserts, sterile
description
24 well plate inserts, non-sterile
description
6 well plate inserts with 1.0 μm polycarbonate filter, sterile
description
96 well plate inserts, 8-well strips, sterile
manufacturer/tradename
Scaffdex C00007S
manufacturer/tradename
Scaffdex C00006N
manufacturer/tradename
Scaffdex C10003F
manufacturer/tradename
Scaffdex C00005S
sterility
sterile; γ-irradiated
sterility
non-sterile
sterility
sterile
sterility
sterile
material
polycarbonate , clear polycarbonate
material
polycarbonate , transparent wells, clear polycarbonate
material
clear polycarbonate
material
colorless wells, polycarbonate (medical grade)
feature
lid
feature
lid
feature
lid
feature
lid
size
12 well
size
24 well
size
6 wells
size
96 wells
packaging
pack of 6 ea
packaging
pack of 12 ea
packaging
pack of 3 ea
packaging
pack of 6 ea
Still not finding the right product?
Explore all of our products under CellCrown™ inserts
Hier finden Sie alle aktuellen Versionen:
Analysenzertifikate (COA)