Fortfahren mit
Z708593
Hexagonal weighing boats
size L
Synonym(e):
weighboat, weiging dish
Anmelden zur Ansicht der Organisations- und Vertragspreise.
Größe auswählen
Ansicht ändern
Größe/SKUVerfügbarkeitPreis
Über diesen Artikel
NACRES:
NB.71
UNSPSC Code:
41121812
Material:
hexagonal , polystyrene
Packaging:
pack of 500 ea
Manufacturer/tradename:
Heathrow Scientific HS14251C
Preise und Verfügbarkeit sind derzeit nicht verfügbar.
Technischer Dienst
Benötigen Sie Hilfe? Unser Team von erfahrenen Wissenschaftlern ist für Sie da.
Unterstützung erhalten
material
hexagonal , polystyrene
size
L
feature
antistatic
packaging
pack of 500 ea
manufacturer/tradename
Heathrow Scientific HS14251C
bottom I.D.
3 in. (76 mm)
top I.D.
4 in. (102 mm)
General description
  • Hexagonal weighing boats are ideal for handling samples for weighing.
  • Flexible boats can be easily bent to form pour spouts for spill-free transfer and prevent sample loss.
  • Flat bottoms and sloping slides provide an accurate transfer.
  • These antistatic, polystyrene boats are non-reactive to most substances.
  • Chemically resistant to most chemicals including weak acids, aqueous solutions, alcohols, and bases.
  • Compatible with microwave moisture analyzers.
  • Resist temperatures to 158 °F (70 °C).
Features and Benefits
  • Available in four different sizes and 1 color
  • Pack of 500
  • FDA/ food grade compliant products
  • Disposable
Ähnliche Artikel vergleichen
Vollständigen Vergleich anzeigen
Unterschiede anzeigen
1 of 1
Dieser Artikel
Z708577Z708615HS120882
description
size L
description
size S
description
size XL
description
size M, anti-static, sterile; electron beam irradiated
manufacturer/tradename
Heathrow Scientific HS14251C
manufacturer/tradename
Heathrow Scientific HS14251A
manufacturer/tradename
Heathrow Scientific HS14251D
manufacturer/tradename
Heathrow Scientific 120882
material
polystyrene , hexagonal
material
polystyrene , hexagonal
material
polystyrene , hexagonal
material
hexagonal white polystyrene
packaging
pack of 500 ea
packaging
pack of 500 ea
packaging
pack of 500 ea
packaging
pack of 200 ea (40 x packs of 5 ea)
top I.D.
4 in. (102 mm)
top I.D.
1.5 in. (38 mm)
top I.D.
5 in. (127 mm)
top I.D.
-
size
L
size
S
size
XL
size
M
bottom I.D.
3 in. (76 mm)
bottom I.D.
1 in. (25 mm)
bottom I.D.
3.5 in. (89 mm)
bottom I.D.
-
Still not finding the right product?
Explore all of our products under Hexagonal weighing boats
Hier finden Sie alle aktuellen Versionen:
Analysenzertifikate (COA)