Fortfahren mit
Z707708
TPP® centrifuge tubes
capacity 50 mL, polypropylene, conical bottom tube
Synonym(e):
TPP Centrifuge Tubes, TPP 91050, sample collection tube, sample receptacle, sample tube
Anmelden zur Ansicht der Organisations- und Vertragspreise.
Größe auswählen
Ansicht ändern
Größe/SKUVerfügbarkeitPreis
Über diesen Artikel
NACRES:
NB.11
UNSPSC Code:
41121703
Capacity:
50 mL
Material:
cap, conical bottom tube, polypropylene , transparent tube
Sterility:
sterile
Packaging:
pack of 360 ea
Feature:
RCF 15,500 × g
Preise und Verfügbarkeit sind derzeit nicht verfügbar.
Technischer Dienst
Benötigen Sie Hilfe? Unser Team von erfahrenen Wissenschaftlern ist für Sie da.
Unterstützung erhalten
material
cap, conical bottom tube, polypropylene , transparent tube
sterility
sterile
feature
RCF 15,500 × g
packaging
pack of 360 ea
manufacturer/tradename
TPP 91050
capacity
50 mL
General description
The TPP 50 mL and 15 mL Centrifuge Tubes are compatible with all standard centrifuges and their innovative design focuses on a tight seal, excellent transparency, clear graduations, and highly visible marking areas. The yellow screw cap closure on the TPP Centrifuge Tubes is tight and protected against overturning. The transparency of the tubes is excellent and the white marking areas on the top of the cap and side of the tube are highly visible. On the conical tubes, the distinct white graduations are continued to the bottom of the tube to accommodate small sample volumes. On the 500 mL tubes, the scale starts at 0.5 mL, and on the 15 mL tubes, the scale starts at 0.1mL.
Temperature Range for Polypropylene Tubes: -196°C to +121°C; Polystyrene Tubes: -40°C to +70°C; Maximum RCF for Polypropylene Tubes: 15500 × g*; Polystyrene Tubes: 1700 × g*.
* Depending on adapter and temperature
All Tubes are gamma Irradiated
Legal Information
TPP is a registered trademark of TPP Techno Plastic Products AG
Ähnliche Artikel vergleichen
Vollständigen Vergleich anzeigen
Unterschiede anzeigen
1 of 1
Dieser Artikel
Z707716Z707732CLS4558
description
capacity 50 mL, polypropylene, conical bottom tube
description
capacity 50 mL, polypropylene
description
capacity 15 mL, polystyrene, conical
description
50 mL centrifuge tubes, polypropylene, conical bottom w/ CentriStar cap, universal rack packed, sterile, natural, 25/universal rack, 300/cs
sterility
sterile
sterility
sterile
sterility
sterile
sterility
sterile
manufacturer/tradename
TPP 91050
manufacturer/tradename
TPP 91051
manufacturer/tradename
TPP 91115
manufacturer/tradename
Corning 4558
material
conical bottom tube, polypropylene , cap, transparent tube
material
transparent tube, cap, polypropylene , conical bottom tube (with rim)
material
cap, polystyrene , conical bottom tube, transparent tube
material
conical bottom tube, polypropylene , cap (CentriStar™), clear tube
capacity
50 mL
capacity
50 mL
capacity
15 mL
capacity
50 mL
packaging
pack of 360 ea
packaging
pack of 320 ea
packaging
pack of 800 ea
packaging
pack of 25, case of 300
feature
RCF 15,500 × g
feature
-
feature
RCF 1,700 × g
feature
RCF 17,000 × g, disposable, self-standing: no, graduations 5 ‑ 50 mL graduation interval: 2.5 mL, RNase and DNase free, RCF 15500 × g
Still not finding the right product?
Explore all of our products under TPP® centrifuge tubes
Hier finden Sie alle aktuellen Versionen:
Analysenzertifikate (COA)