Fortfahren mit
Z694517
Nanofiber multiwell plate inserts, with random oriented nanofibers
for 12 well plates
Synonym(e):
3D, 3D Cell Culture, 3d plates, tissue culture plates, Nanofiber Solutions 121201
Anmelden zur Ansicht der Organisations- und Vertragspreise.
Größe auswählen
Ansicht ändern
Größe/SKUVerfügbarkeitPreis
12 ea
Warenkorb auf Verfügbarkeit prüfen
€ 107,00
Über diesen Artikel
NACRES:
NB.22
UNSPSC Code:
41122107
Material:
(polycaprolactone (PCL)), flat clear wells, transparent polycaprolactone (PCL)
Pore size:
700 nm (Fiber spacing)
Sterility:
sterile; electron beam irradiated, sterile
Binding type:
(plasma surface treated)
€ 107,00
Warenkorb auf Verfügbarkeit prüfen
​
Technischer Dienst
Benötigen Sie Hilfe? Unser Team von erfahrenen Wissenschaftlern ist für Sie da.
Unterstützung erhalten
material
(polycaprolactone (PCL)), flat clear wells, transparent polycaprolactone (PCL)
sterility
sterile; electron beam irradiated, sterile
feature
lid
packaging
pack of 12 ea
manufacturer/tradename
Nanofiber Solutions 121201
size
12 wells
pore size
700 nm (Fiber spacing)
suitability
suitable for (Tissue Engineering, Cancer Research)
application(s)
3D cell culture
binding type
(plasma surface treated)
General description
Nanofiber cell culture dishes, multiwell plates and well plate inserts combine the best of 3D and 2D cell culture. The nanofibers are optically transparent to allow for live-cell imaging and real time quantification of cell mobility. The nanofibers mimic the 3D topography in vivo, and can be easily coated with ECM proteins for cancer research. The cells can be simply removed for protein or gene analysis.
The nanofibers have higher expansion rates, better differentiation control, and has a synthetic surface with no animal contamination, ideal for stem cells research and immunohistochemistry staining. Nanofiber scaffolds can be easily shaped or sized, made from nearly any synthetic or natural polymer for customized tissue engineering. They can then be implanted in vivo or used in vitro. The standard plate dimensions available will work with current automated equipment for high throughput drug discovery needs.
The cell culture dishes, plates and inserts are available with either randomly oriented fibers or aligned fibers.
Ähnliche Artikel vergleichen
Vollständigen Vergleich anzeigen
Unterschiede anzeigen
1 of 1
Dieser Artikel
Z694509Z694614Z694444
description
for 12 well plates
description
for 6 well plates
description
for 12 well plates
description
size 24 wells
manufacturer/tradename
Nanofiber Solutions 121201
manufacturer/tradename
Nanofiber Solutions 60601
manufacturer/tradename
Nanofiber Solutions 121202
manufacturer/tradename
Nanofiber Solutions 2401
binding type
(plasma surface treated)
binding type
(plasma surface treated)
binding type
(plasma surface treated)
binding type
non-treated surface
pore size
700 nm (Fiber spacing)
pore size
700 nm (Fiber spacing)
pore size
700 nm (Fiber spacing)
pore size
-
sterility
sterile; electron beam irradiated
sterility
sterile, sterile; electron beam irradiated
sterility
sterile, sterile; electron beam irradiated
sterility
sterile
material
flat clear wells, (polycaprolactone (PCL)), transparent polycaprolactone (PCL)
material
(polycaprolactone (PCL)), transparent polycaprolactone (PCL) , flat clear wells
material
transparent wells, transparent polycaprolactone (PCL)
material
transparent polycaprolactone (PCL) , flat bottom clear wells, (polycaprolactone (PCL))
suitability
suitable for (Tissue Engineering, Cancer Research)
suitability
suitable for (Tissue Engineering, Cancer Research)
suitability
suitable for (Tissue Engineering, Cancer Research)
suitability
suitable for (Tissue engineering, cancer research)
Still not finding the right product?
Hier finden Sie alle aktuellen Versionen:
Analysenzertifikate (COA)