Fortfahren mit
Z694444
Nanofiber multiwell plate, with random oriented nanofibers
size 24 wells
Synonym(e):
24 well culture plates, 3D, 3D Cell Culture, 3d plates, tissue culture plates, Nanofiber Solutions 2401
Anmelden zur Ansicht der Organisations- und Vertragspreise.
Größe auswählen
Ansicht ändern
Größe/SKUVerfügbarkeitPreis
1 ea
Warenkorb auf Verfügbarkeit prüfen
€ 97,10
Über diesen Artikel
NACRES:
NB.22
UNSPSC Code:
41121800
Material:
(polycaprolactone (PCL)), flat bottom clear wells, transparent polycaprolactone (PCL)
Size:
24 wells
Sterility:
sterile
Binding type:
non-treated surface
Feature:
lid
€ 97,10
Warenkorb auf Verfügbarkeit prüfen
Technischer Dienst
Benötigen Sie Hilfe? Unser Team von erfahrenen Wissenschaftlern ist für Sie da.
Unterstützung erhalten
material
(polycaprolactone (PCL)), flat bottom clear wells, transparent polycaprolactone (PCL)
sterility
sterile
feature
lid
manufacturer/tradename
Nanofiber Solutions 2401
size
24 wells
suitability
suitable for (Tissue engineering, cancer research)
binding type
non-treated surface
General description
Nanofiber cell culture dishes, multiwell plates and well plate inserts combine the best of 3D and 2D cell culture. The nanofibers are optically transparent to allow for live-cell imaging and real time quantification of cell mobility. The nanofibers mimic the 3D topography in vivo, and can be easily coated with ECM proteins for cancer research. The cells can be simply removed for protein or gene analysis.
The nanofibers have higher expansion rates, better differentiation control, and has a synthetic surface with no animal contamination, ideal for stem cells research and immunohistochemistry staining. Nanofiber scaffolds can be easily shaped or sized, made from nearly any synthetic or natural polymer for customized tissue engineering. They can then be implanted in vivo or used in vitro. The standard plate dimensions available will work with current automated equipment for high throughput drug discovery needs.
The cell culture dishes, plates and inserts are available with either randomly oriented fibers or aligned fibers.
For fluorescent imaging applications, we recommend using the Nanofiber Solutions chamber slides.
Ähnliche Artikel vergleichen
Vollständigen Vergleich anzeigen
Unterschiede anzeigen
1 of 1
Dieser Artikel
Z694533Z759341Z694568
description
size 24 wells
description
size 24 wells
description
size 6 wells
description
size 384 wells
manufacturer/tradename
Nanofiber Solutions 2401
manufacturer/tradename
Nanofiber Solutions 2402
manufacturer/tradename
Nanofiber Solutions 601
manufacturer/tradename
Nanofiber Solutions 38402
feature
lid
feature
lid
feature
lid
feature
lid
sterility
sterile
sterility
sterile
sterility
sterile
sterility
sterile
binding type
non-treated surface
binding type
non-treated surface
binding type
non-treated surface
binding type
non-treated surface
material
transparent polycaprolactone (PCL) , (polycaprolactone (PCL)), flat bottom clear wells
material
transparent polycaprolactone (PCL) , flat clear wells, (polycaprolactone (PCL))
material
(polycaprolactone (PCL)), flat bottom clear wells, transparent polycaprolactone (PCL)
material
(polycaprolactone (PCL)), flat bottom clear wells, transparent polycaprolactone (PCL)
size
24 wells
size
24 wells
size
6 wells
size
384 wells
Still not finding the right product?
Hier finden Sie alle aktuellen Versionen:
Analysenzertifikate (COA)