Fortfahren mit
Z694460
Nanofiber multiwell plate, with random oriented nanofibers
size 384 wells
Synonym(e):
384 multiwell plates, 384 well microplates, 384 well microtiter plates, 384 well plates, 3D, 3D Cell Culture, 3d plates, tissue culture plates, Nanofiber Solutions 38401
Anmelden zur Ansicht der Organisations- und Vertragspreise.
Größe auswählen
Ansicht ändern
Größe/SKUVerfügbarkeitPreis
1 ea
Warenkorb auf Verfügbarkeit prüfen
€ 101,00
Über diesen Artikel
NACRES:
NB.13
UNSPSC Code:
41121800
Material:
(polycaprolactone (PCL)), flat bottom clear wells, transparent polycaprolactone (PCL)
Size:
384 wells
Sterility:
sterile
Binding type:
non-treated surface
Feature:
lid
€ 101,00
Warenkorb auf Verfügbarkeit prüfen
​
Technischer Dienst
Benötigen Sie Hilfe? Unser Team von erfahrenen Wissenschaftlern ist für Sie da.
Unterstützung erhalten
material
(polycaprolactone (PCL)), flat bottom clear wells, transparent polycaprolactone (PCL)
sterility
sterile
feature
lid
manufacturer/tradename
Nanofiber Solutions 38401
technique(s)
cell culture: suitable
size
384 wells
suitability
suitable for (Tissue Engineering, Cancer Research)
application(s)
3D cell culture
binding type
non-treated surface
General description
Nanofiber cell culture dishes, multiwell plates and well plate inserts combine the best of 3D and 2D cell culture. The nanofibers are optically transparent to allow for live-cell imaging and real time quantification of cell mobility. The nanofibers mimic the 3D topography in vivo, and can be easily coated with ECM proteins for cancer research. The cells can be simply removed for protein or gene analysis.
The nanofibers have higher expansion rates, better differentiation control, and has a synthetic surface with no animal contamination, ideal for stem cells research and immunohistochemistry staining. Nanofiber scaffolds can be easily shaped or sized, made from nearly any synthetic or natural polymer for customized tissue engineering. They can then be implanted in vivo or used in vitro. The standard plate dimensions available will work with current automated equipment for high throughput drug discovery needs.
The cell culture dishes, plates and inserts are available with either randomly oriented fibers or aligned fibers.
For fluorescent imaging applications, we recommend using the Nanofiber Solutions chamber slides.
Ähnliche Artikel vergleichen
Vollständigen Vergleich anzeigen
Unterschiede anzeigen
1 of 1
Dieser Artikel
Z759341Z694452Z694444
description
size 384 wells
description
size 6 wells
description
size 96 wells
description
size 24 wells
sterility
sterile
sterility
sterile
sterility
sterile
sterility
sterile
manufacturer/tradename
Nanofiber Solutions 38401
manufacturer/tradename
Nanofiber Solutions 601
manufacturer/tradename
Nanofiber Solutions 9601
manufacturer/tradename
Nanofiber Solutions 2401
binding type
non-treated surface
binding type
non-treated surface
binding type
non-treated surface
binding type
non-treated surface
feature
lid
feature
lid
feature
lid
feature
lid
material
(polycaprolactone (PCL)), flat bottom clear wells, transparent polycaprolactone (PCL)
material
transparent polycaprolactone (PCL) , (polycaprolactone (PCL)), flat bottom clear wells
material
transparent polycaprolactone (PCL) , flat clear wells, (polycaprolactone (PCL))
material
transparent polycaprolactone (PCL) , (polycaprolactone (PCL)), flat bottom clear wells
suitability
suitable for (Tissue Engineering, Cancer Research)
suitability
suitable for (Tissue Engineering, Cancer Research)
suitability
suitable for (Tissue Engineering, Cancer Research)
suitability
suitable for (Tissue engineering, cancer research)
Still not finding the right product?
Hier finden Sie alle aktuellen Versionen:
Analysenzertifikate (COA)