Fortfahren mit
Z364592
Nalgene® media bottles, style 2020
capacity 1000 mL
Synonym(e):
bottle nalgene, nalgene containers, sterile bottles, Nalgene 342020-1000, media container, reagent bottle
Anmelden zur Ansicht der Organisations- und Vertragspreise.
Größe auswählen
Ansicht ändern
Größe/SKUVerfügbarkeitPreis
Über diesen Artikel
UNSPSC Code:
41121800
Capacity:
1000 mL
Material:
high-density polyethylene cap, square bottle, transparent PETG
Sterility:
sterile; γ-irradiated
Feature:
graduations: no, with cap
Packaging:
pack of 24 ea (2 xpack of 12)
Preise und Verfügbarkeit sind derzeit nicht verfügbar.
Technischer Dienst
Benötigen Sie Hilfe? Unser Team von erfahrenen Wissenschaftlern ist für Sie da.
Unterstützung erhalten
material
high-density polyethylene cap, square bottle, transparent PETG
joint
Joints threaded neck (38-430mm)
description
Not autoclavable
sterility
sterile; γ-irradiated
feature
graduations: no, with cap
packaging
pack of 24 ea (2 xpack of 12)
manufacturer/tradename
Nalgene® 342020-1000
capacity
1000 mL
General description
Media bottles (square). Same materials and design as Style 2019, but without shrink band around neck.
Features and Benefits
  • Extensive validation documentation is available
  • Produced from durable, break-resistant polyethylene terephthalate copolyester, glycol modified (PETG) that enhance reduced permeability to CO2/O2.
  • Tamper evident shrink sleeves used along with moulded in graduations and neck ring.
  • Non-pyrogenic and non-cytotoxic
  • Animal derived component free (ADCF) material for bottle and closure material.
  • Leakproof and transparent
  • Devoid of costly washing, depyrogenation steps
  • Prolonged storage of pH-sensitive liquids such as culture media
Legal Information
Nalgene is a registered trademark of Thermo Fisher Scientific or its subsidiaries
Ähnliche Artikel vergleichen
Vollständigen Vergleich anzeigen
Unterschiede anzeigen
1 of 1
Dieser Artikel
Z741028Z364576Z364568
description
capacity 1000 mL
description
capacity 2000 mL
description
capacity 500 mL
description
capacity 250 mL
sterility
sterile; γ-irradiated
sterility
sterile; γ-irradiated
sterility
sterile; γ-irradiated
sterility
sterile; γ-irradiated
feature
with cap, graduations: no
feature
graduations: no, with cap
feature
graduations: no, with cap
feature
with cap, graduations: no
capacity
1000 mL
capacity
2000 mL
capacity
500 mL
capacity
250 mL
material
square bottle, high-density polyethylene cap, transparent PETG
material
square bottle, high-density polyethylene cap, transparent PETG
material
transparent PETG , square bottle, high-density polyethylene cap
material
square bottle, high-density polyethylene cap, transparent PETG
packaging
pack of 24 ea (2 xpack of 12)
packaging
pack of 12 ea (2 xpack of 6)
packaging
pack of 40 ea (2 x pack of 20)
packaging
pack of 60 ea (2 xpack of 30)
joint
Joints threaded neck (38-430mm)
joint
Joints threaded neck (53B)
joint
Joints threaded neck (38-430mm)
joint
Joints threaded neck (38-430mm)
Still not finding the right product?
Hier finden Sie alle aktuellen Versionen:
Analysenzertifikate (COA)