Fortfahren mit
Z364584
Nalgene® media bottles, style 2020
capacity 650 mL
Aktionsangebot verfügbar
Synonym(e):
bottle nalgene, nalgene containers, sterile bottles, Nalgene 342020-9500, media container, reagent bottle
Anmelden zur Ansicht der Organisations- und Vertragspreise.
Spend € 450 — Save 5 %; Spend € 700 — Save 10 %; Spend € 1.500 — Save 15 %*
Anmelden zur Überprüfung der Spielberechtigung. Produkte kaufen
Größe auswählen
Ansicht ändern
Größe/SKUVerfügbarkeitPreis
48 ea
Warenkorb auf Verfügbarkeit prüfen
€ 313,00
Über diesen Artikel
NACRES:
NB.17
UNSPSC Code:
41121800
Capacity:
650 mL
Material:
high-density polyethylene cap, square bottle, transparent PETG
Sterility:
sterile; γ-irradiated
Feature:
graduations: no, with cap
Packaging:
pack of 48 ea (2 x pack of 24)
€ 313,00
Sparen Sie bis zu 15 %
Warenkorb auf Verfügbarkeit prüfen
​
Technischer Dienst
Benötigen Sie Hilfe? Unser Team von erfahrenen Wissenschaftlern ist für Sie da.
Unterstützung erhalten
material
high-density polyethylene cap, square bottle, transparent PETG
joint
Joints threaded neck (38-430mm)
description
Not autoclavable
sterility
sterile; γ-irradiated
feature
graduations: no, with cap
packaging
pack of 48 ea (2 x pack of 24)
manufacturer/tradename
Nalgene® 342020-9500
capacity
650 mL
General description
Media bottles (square). Same materials and design as Style 2019, but without shrink band around neck.
Features and Benefits
  • Extensive validation documentation is available
  • Produced from durable, break-resistant polyethylene terephthalate copolyester, glycol modified (PETG) that enhance reduced permeability to CO2/O2.
  • Tamper evident shrink sleeves used along with moulded in graduations and neck ring.
  • Non-pyrogenic and non-cytotoxic
  • Animal derived component free (ADCF) material for bottle and closure material.
  • Leakproof and transparent
  • Devoid of costly washing, depyrogenation steps
  • Prolonged storage of pH-sensitive liquids such as culture media
Legal Information
Nalgene is a registered trademark of Thermo Fisher Scientific or its subsidiaries
Ähnliche Artikel vergleichen
Vollständigen Vergleich anzeigen
Unterschiede anzeigen
1 of 1
Dieser Artikel
Z364568Z364541Z364533
description
capacity 650 mL
description
capacity 250 mL
description
capacity 125 mL
description
capacity 60 mL
capacity
650 mL
capacity
250 mL
capacity
125 mL
capacity
60 mL
feature
graduations: no, with cap
feature
with cap, graduations: no
feature
graduations: no, with cap
feature
graduations: no, with cap
sterility
sterile; γ-irradiated
sterility
sterile; γ-irradiated
sterility
sterile; γ-irradiated
sterility
sterile; γ-irradiated
material
high-density polyethylene cap, square bottle, transparent PETG
material
square bottle, high-density polyethylene cap, transparent PETG
material
high-density polyethylene cap, square bottle, transparent PETG
material
square bottle, transparent PETG , high-density polyethylene cap
packaging
pack of 48 ea (2 x pack of 24)
packaging
pack of 60 ea (2 xpack of 30)
packaging
pack of 96 ea (4 xpack of 24)
packaging
pack of 200 ea (5 xpack of 40)
manufacturer/tradename
Nalgene® 342020-9500
manufacturer/tradename
Nalgene® 342020-0250
manufacturer/tradename
Nalgene® 342020-0125
manufacturer/tradename
Nalgene® 342020-0060
Still not finding the right product?
Hier finden Sie alle aktuellen Versionen:
Analysenzertifikate (COA)