Fortfahren mit
Z328871
BRAND® Magnetrührstäbchen, PTFE, zylindrisch
L 60 mm, bar diam. 9 mm
Synonym(e):
cylindrical stir bar, magnetic bar, magnetic stir bar, spinbar, stir bar
Anmelden zur Ansicht der Organisations- und Vertragspreise.
Größe auswählen
Ansicht ändern
Größe/SKUVerfügbarkeitPreis
10 ea
Warenkorb auf Verfügbarkeit prüfen
€ 107,00
Über diesen Artikel
NACRES:
NB.72
UNSPSC Code:
41103806
Material:
PTFE-coated , cylindrical
Manufacturer/tradename:
BRAND 137140
€ 107,00
Warenkorb auf Verfügbarkeit prüfen
Technischer Dienst
Benötigen Sie Hilfe? Unser Team von erfahrenen Wissenschaftlern ist für Sie da.
Unterstützung erhalten
Quality Segment
material
PTFE-coated , cylindrical
manufacturer/tradename
BRAND 137140
L
60 mm
bar diam.
9 mm
General description
BRAND® magnetic cylindrical stirring bars are expertly designed for optimal mixing performance, featuring a smooth, round shape that enhances stirring efficiency. Their Alnico V magnetic cores are protected by a robust PTFE (polytetrafluoroethylene) coating, ensuring durability and resistance to chemical damage. These stirring bars are particularly effective for use in large volumes or thick mixtures, making them ideal for a variety of laboratory applications.
Features and Benefits
  • BRAND® magnetic cylindrical stirring bars are round with a smooth surface.
  • The Alnico V magnetic cores are encapsulated with high-grade, crack-resistant PTFE (polytetrafluoroethylene) coating.
  • Ideal for large volumes or highly viscous mixtures.
Other Notes
BR137140 (India)
Legal Information
BRAND is a registered trademark of BRAND GMBH + CO KG
Ähnliche Artikel vergleichen
Vollständigen Vergleich anzeigen
Unterschiede anzeigen
1 of 1
Dieser Artikel
Z328812Z328944Z328715
description
L 60 mm, bar diam. 9 mm
description
L 40 mm, bar diam. 8 mm
description
L 80 mm, bar diam. 9 mm
description
L 15 mm, bar diam. 4.5 mm
manufacturer/tradename
BRAND 137140
manufacturer/tradename
BRAND 137130
manufacturer/tradename
BRAND 137150
manufacturer/tradename
BRAND 137110
material
cylindrical , PTFE-coated
material
PTFE-coated , cylindrical
material
PTFE-coated , cylindrical
material
cylindrical , PTFE-coated
bar diam.
9 mm
bar diam.
8 mm
bar diam.
9 mm
bar diam.
4.5 mm
Quality Level
200
Quality Level
200
Quality Level
-
Quality Level
200
L
60 mm
L
40 mm
L
80 mm
L
15 mm
Still not finding the right product?
Hier finden Sie alle aktuellen Versionen:
Analysenzertifikate (COA)