Fortfahren mit
Z328944
BRAND® Magnetrührstäbchen, PTFE, zylindrisch
L 80 mm, bar diam. 9 mm
Synonym(e):
cylindrical stir bar, magnetic bar, magnetic stir bar, spinbar, stir bar
Anmelden zur Ansicht der Organisations- und Vertragspreise.
Größe auswählen
Ansicht ändern
Größe/SKUVerfügbarkeitPreis
10 ea
Warenkorb auf Verfügbarkeit prüfen
€ 116,00
Über diesen Artikel
NACRES:
NB.72
UNSPSC Code:
41103806
Material:
PTFE-coated , cylindrical
Manufacturer/tradename:
BRAND 137150
€ 116,00
Warenkorb auf Verfügbarkeit prüfen
​
Technischer Dienst
Benötigen Sie Hilfe? Unser Team von erfahrenen Wissenschaftlern ist für Sie da.
Unterstützung erhalten
material
PTFE-coated , cylindrical
manufacturer/tradename
BRAND 137150
L
80 mm
bar diam.
9 mm
General description
BRAND® magnetic cylindrical stirring bars are expertly designed for optimal mixing performance, featuring a smooth, round shape that enhances stirring efficiency. Their Alnico V magnetic cores are protected by a robust PTFE (polytetrafluoroethylene) coating, ensuring durability and resistance to chemical damage. These stirring bars are particularly effective for use in large volumes or thick mixtures, making them ideal for a variety of laboratory applications.
Features and Benefits
  • BRAND® magnetic cylindrical stirring bars are round with a smooth surface.
  • The Alnico V magnetic cores are encapsulated with high-grade, crack-resistant PTFE (polytetrafluoroethylene) coating.
  • Ideal for large volumes or highly viscous mixtures.
Other Notes
BR137150 (India)
Legal Information
BRAND is a registered trademark of BRAND GMBH + CO KG
Ähnliche Artikel vergleichen
Vollständigen Vergleich anzeigen
Unterschiede anzeigen
1 of 1
Dieser Artikel
Z328715Z328936Z328898
description
L 80 mm, bar diam. 9 mm
description
L 15 mm, bar diam. 4.5 mm
description
L 8 mm, bar diam. 3 mm
description
L 7 mm, bar diam. 2 mm
manufacturer/tradename
BRAND 137150
manufacturer/tradename
BRAND 137110
manufacturer/tradename
BRAND 137104
manufacturer/tradename
BRAND 137106
material
PTFE-coated , cylindrical
material
cylindrical , PTFE-coated
material
PTFE-coated , cylindrical
material
cylindrical , PTFE-coated
L
80 mm
L
15 mm
L
8 mm
L
7 mm
bar diam.
9 mm
bar diam.
4.5 mm
bar diam.
3 mm
bar diam.
2 mm
Still not finding the right product?
Hier finden Sie alle aktuellen Versionen:
Analysenzertifikate (COA)