Größe auswählen
| Ihnen/SKU | Verfügbarkeit | Preis |
|---|---|---|
1 pkg | Warenkorb auf Verfügbarkeit prüfen | € 27,50 |
Über diesen Artikel
€ 27,50
material
polypropylene
sterility
non-sterile
feature
color-coded red, DE-M marking
packaging
pack of 10 ea
manufacturer/tradename
BRAND 702858, BRAND
maximum volume
5 mL
volume
1-5 mL
Other Notes
Legal Information
1 of 1
Dieser Artikel | |||
|---|---|---|---|
| description volume 1-5 mL | description volume 2-10 mL | description volume 200-1000 μL | description volume 1-5 μL |
| manufacturer/tradename BRAND, BRAND 702858 | manufacturer/tradename BRAND 702860 | manufacturer/tradename BRAND, BRAND 702854 | manufacturer/tradename BRAND 701900 |
| volume 1-5 mL | volume 2-10 mL | volume 200-1000 μL | volume 1-5 μL |
| feature color-coded red, DE-M marking | feature DE-M marking, color-coded orange | feature DE-M marking, color-coded yellow | feature color-coded white, DE-M marking |
| sterility non-sterile | sterility non-sterile | sterility non-sterile | sterility non-sterile |
| material polypropylene | material polypropylene | material polypropylene | material glass |
| packaging pack of 10 ea | packaging pack of 10 ea | packaging pack of 10 ea | packaging pack of 100 ea |
Still not finding the right product?
Explore all of our products under BRAND® Transferpettor Verschlüsse
Hier finden Sie alle aktuellen Versionen:
Besitzen Sie dieses Produkt bereits?