Fortfahren mit
Z327077
BRAND® BLAUBRAND® volumetric flask, glass stopper, amber glass
volume 200 mL, accuracy: 0.15 mL
Synonym(e):
amber flask, analytical flask, calibration flask with stopper, measuring flask, volumetric flask
Anmelden zur Ansicht der Organisations- und Vertragspreise.
Größe auswählen
Ansicht ändern
Größe/SKUVerfügbarkeitPreis
2 ea
Warenkorb auf Verfügbarkeit prüfen
€ 97,90
Über diesen Artikel
NACRES:
NB.43
UNSPSC Code:
41121804
Capacity:
200 mL
Material:
borosilicate glass 3.3 flask
Packaging:
pack of 2 ea
€ 97,90
Warenkorb auf Verfügbarkeit prüfen
​
Technischer Dienst
Benötigen Sie Hilfe? Unser Team von erfahrenen Wissenschaftlern ist für Sie da.
Unterstützung erhalten
material
borosilicate glass 3.3 flask
joint
joint ST/NS neck14/23
feature
class A DE-M, DIN EN ISO 1042. Calibrated to contain (TC, In).
packaging
pack of 2 ea
manufacturer/tradename
BRAND 37450
volume measuring range
200 mL, accuracy: 0.15 mL
capacity
200 mL
General description
Volumetric flasks according to DIN EN ISO 1042, Class A, conformity certified. Made from Boro 3.3, calibrated to contain (TC, IN). Supplied with a interchangeable amber glass stopper. Marks and inscriptions in high contrast white enamel.
All BLAUBRAND® volumetric flasks are supplied with a batch number and an accompanying batch certificate in the original packaging.
Other Notes
BR37450 (India)
Legal Information
BLAUBRAND is a registered trademark of BRAND GMBH + CO KG
BRAND is a registered trademark of BRAND GMBH + CO KG
Ähnliche Artikel vergleichen
Vollständigen Vergleich anzeigen
Unterschiede anzeigen
1 of 1
Dieser Artikel
Z327085Z327093Z326992
description
volume 200 mL, accuracy: 0.15 mL
description
volume 250 mL, accuracy: 0.15 mL
description
volume 500 mL, accuracy: 0.25 mL
description
volume 5 mL, accuracy: 0.08 mL
manufacturer/tradename
BRAND 37450
manufacturer/tradename
BRAND 37451
manufacturer/tradename
BRAND 37452
manufacturer/tradename
BRAND 37438
capacity
200 mL
capacity
250 mL
capacity
500 mL
capacity
5 mL
feature
class A DE-M, DIN EN ISO 1042. Calibrated to contain (TC, In).
feature
class A DE-M, DIN EN ISO 1042. Calibrated to contain (TC, In).
feature
class A DE-M, DIN EN ISO 1042. Calibrated to contain (TC, In).
feature
class A DE-M, DIN EN ISO 1042. Calibrated to contain (TC, In).
joint
joint ST/NS neck14/23
joint
joint ST/NS neck14/23
joint
joint ST/NS neck19/26
joint
joint ST/NS neck10/19
material
borosilicate glass 3.3 flask
material
borosilicate glass 3.3 flask
material
borosilicate glass 3.3 flask
material
borosilicate glass 3.3 flask, wide-neck
packaging
pack of 2 ea
packaging
pack of 2 ea
packaging
pack of 2 ea
packaging
pack of 2 ea
Still not finding the right product?
Hier finden Sie alle aktuellen Versionen:
Analysenzertifikate (COA)