Größe auswählen
| Größe/SKU | Verfügbarkeit | Preis |
|---|---|---|
2 ea | Warenkorb auf Verfügbarkeit prüfen | € 65,40 |
Über diesen Artikel
€ 65,40
material
borosilicate glass 3.3 flask
Quality Segment
joint
joint ST/NS neck10/19
description
calibrated to contain (TC, IN)
feature
class A DE-M, DIN EN ISO 1042. Calibrated to contain (TC, In).
packaging
pack of 2 ea
manufacturer/tradename
BRAND 37404
volume measuring range
25 mL, accuracy: 0.04 mL
capacity
25 mL
General description
Other Notes
Legal Information
1 of 1
Dieser Artikel | |||
|---|---|---|---|
| description volume 25 mL, accuracy: 0.04 mL | description volume 50 mL, accuracy: 0.06 mL | description volume 200 mL, accuracy: 0.15 mL | description polypropylene stopper, amber borosilicate glass 3.3 flask, capacity 25 mL, accuracy, 0.06 mL |
| manufacturer/tradename BRAND 37404 | manufacturer/tradename BRAND 37405 | manufacturer/tradename BRAND 37408 | manufacturer/tradename BRAND 37413 |
| capacity 25 mL | capacity 50 mL | capacity 200 mL | capacity 25 mL |
| feature class A DE-M, DIN EN ISO 1042. Calibrated to contain (TC, In). | feature class A DE-M, DIN EN ISO 1042. Calibrated to contain (TC, In). | feature class A DE-M, DIN EN ISO 1042. Calibrated to contain (TC, In). | feature accuracy 0.06 mL, class A DIN EN ISO 1042. Calibrated to contain (TC, In) |
| material borosilicate glass 3.3 flask | material borosilicate glass 3.3 flask | material borosilicate glass 3.3 flask | material polypropylene stopper, amber borosilicate glass 3.3 flask |
| joint joint ST/NS neck10/19 | joint joint ST/NS neck12/21 | joint joint ST/NS neck14/23 | joint joint ST/NS12/21 |
| packaging pack of 2 ea | packaging pack of 2 ea | packaging pack of 2 ea | packaging pack of 2 ea |
Still not finding the right product?
Explore all of our products under BRAND® BLAUBRAND® Messkolben, PP-Stopfen, Braunglas
Hier finden Sie alle aktuellen Versionen:
Besitzen Sie dieses Produkt bereits?