Größe auswählen
| Größe/SKU | Verfügbarkeit | Preis |
|---|---|---|
12 ea | Warenkorb auf Verfügbarkeit prüfen | € 78,60 |
Über diesen Artikel
€ 78,60
material
AR-Glas®
Quality Segment
description
total delivery Ex + 15 sec waiting time
feature
class AS
packaging
pack of 12 ea
manufacturer/tradename
BRAND 29703
capacity
2 mL
General description
According to DIN EN ISO 648.
Other Notes
Legal Information
1 of 1
Dieser Artikel | |||
|---|---|---|---|
| description capacity 2 mL, one mark | description capacity 0.5 mL, one mark | description capacity 1 mL, one mark | description capacity 2.5 mL, one mark |
| feature class AS | feature class AS | feature class AS | feature class AS |
| manufacturer/tradename BRAND 29703 | manufacturer/tradename BRAND 29701 | manufacturer/tradename BRAND 29702 | manufacturer/tradename BRAND 29704 |
| capacity 2 mL | capacity 0.5 mL | capacity 1 mL | capacity 2.5 mL |
| material AR-Glas® | material AR-Glas® | material AR-Glas® | material AR-Glas® |
| packaging pack of 12 ea | packaging pack of 12 ea | packaging pack of 12 ea | packaging pack of 6 ea |
| description total delivery Ex + 15 sec waiting time | description total delivery Ex + 15 sec waiting time | description total delivery Ex + 15 sec waiting time | description total delivery Ex + 15 sec waiting time |
Still not finding the right product?
Explore all of our products under BRAND® BLAUBRAND® Vollpipette, kalibriert für die Abgabe (TD, EX)
Hier finden Sie alle aktuellen Versionen:
Besitzen Sie dieses Produkt bereits?