Merck
TMO382099-1000
Nalgene® Certified Low Particulate narrow-mouth bottle
natural high-density polyethylene, volume 1000 mL, case of 24 ea
Synonym(e):
HDPE bottle, certified nalgene bottle
Größe auswählen
1 EACH
€ 304,00
€ 304,00
Check Cart for Availability
Größe auswählen
Ansicht ändern
1 EACH
€ 304,00
About This Item
UNSPSC-Code:
41121800
NACRES:
NB.43
€ 304,00
Check Cart for Availability
Materialien
natural high-density polyethylene
Verpackung
case of 24 ea
Hersteller/Markenname
Nalgene 382099-1000
Kapazität
1000 mL
Volumen
1000 mL
Allgemeine Beschreibung
Effectively store high-purity chemicals with Thermo Scientific Nalgene Certified Low Particulate Narrow-Mouth HDPE Bottles with Closure, which are extremely low in trace metal content. Controlled environment manufacturing minimizes particulate contamination.
Rechtliche Hinweise
Nalgene is a registered trademark of Thermo Fisher Scientific or its subsidiaries
Hier finden Sie alle aktuellen Versionen:
Analysenzertifikate (COA)