Merck
TMO382019-0500
Nalgene® Certified Clean bottles
clear PETG, volume 500 mL, case of 24 ea
Synonym(e):
PETG bottle, certified nalgene bottle
Größe auswählen
24 EACH
€ 460,00
€ 460,00
Check Cart for Availability
Größe auswählen
Ansicht ändern
24 EACH
€ 460,00
About This Item
UNSPSC-Code:
41121800
NACRES:
NB.43
€ 460,00
Check Cart for Availability
Materialien
clear PETG
Verpackung
case of 24 ea
Hersteller/Markenname
Nalgene 382019-0500
Kapazität
500 mL
Volumen
500 mL
Allgemeine Beschreibung
Protect against leakage, breakage and radiation contamination in cleanroom applications with Thermo Scientific Nalgene PETG Certified Clean Containers. Suitable for pharmaceutical and biotechnology applications that require processing and storing bulk intermediates and critical reagents such as vaccine and protein therapeutic preparations. A safe alternative for your lab with no sacrifice of accuracy.
Leistungsmerkmale und Vorteile
  • Lot-certified for low particulates, these containers features a unique leakproof† design and are suitable for transport.
  • Exhibit excellent gas barrier properties and ensure pH stability for buffered solutions.
  • Resins are free of animal-derived components.
  • Sterile, tamper-resistant seal ensures product integrity prior to use.
Rechtliche Hinweise
Nalgene is a registered trademark of Thermo Fisher Scientific or its subsidiaries
Hier finden Sie alle aktuellen Versionen:
Analysenzertifikate (COA)