Merck
TMO332189-0016
Nalgene® economy wide-mouth bottle
natural high-density polyethylene, volume 500 mL, case of 125 ea
Synonym(e):
HDPE bottle, nalgene HDPE bottle
Größe auswählen
125 EACH
€ 430,00
€ 430,00
Check Cart for Availability
Größe auswählen
Ansicht ändern
125 EACH
€ 430,00
About This Item
UNSPSC-Code:
41121800
NACRES:
NB.43
€ 430,00
Check Cart for Availability
Materialien
natural high-density polyethylene
Verpackung
case of 125 ea
Hersteller/Markenname
Nalgene 332189-0016
Kapazität
500 mL
Volumen
500 mL
Allgemeine Beschreibung
Perform short-term general purpose laboratory and field applications with Thermo Scientific Nalgene Wide-Mouth HDPE Economy Bottles with Closure. These light-duty translucent bottles are a lightweight version of the original Nalgene bottle, bulk-packed for ultimate economy. Thinner-walled and more flexible than the Nalgene Lab Quality Bottles, they feature a wide mouth for easy filling and offer excellent chemical resistance to most acids, bases and alcohols.
Rechtliche Hinweise
Nalgene is a registered trademark of Thermo Fisher Scientific or its subsidiaries
Hier finden Sie alle aktuellen Versionen:
Analysenzertifikate (COA)