Fortfahren mit
TMO163371
Nunc® cell culture treated flask with solid cap
polystyrene, flask capacity 7 mL, Nunclon® Delta surface, sterile, case of 160 ea
Synonym(e):
cell culture flask, treated flask, Nunc 163371, plastic flask, surface treated flask
Anmelden zur Ansicht der Organisations- und Vertragspreise.
Größe auswählen
Ansicht ändern
Größe/SKUVerfügbarkeitPreis
160 ea
Warenkorb auf Verfügbarkeit prüfen
€ 527,00
Über diesen Artikel
NACRES:
NB.11
UNSPSC Code:
41161502
Material:
polystyrene
Sterility:
sterile
€ 527,00
Warenkorb auf Verfügbarkeit prüfen
Technischer Dienst
Benötigen Sie Hilfe? Unser Team von erfahrenen Wissenschaftlern ist für Sie da.
Unterstützung erhalten
material
polystyrene
sterility
sterile
packaging
case of 160 ea
manufacturer/tradename
Nunc 163371
flask capacity
7 mL
General description
Achieve maximum adhesion for a broad range of cell types with Thermo Scientific Nunc Cell Culture Treated Flasks with Filter Caps. These flasks receive the proprietary Nunclon Delta surface treatment to ensure consistent cell growth. The filter caps allow constant airflow and minimize chances of contamination.
Legal Information
Nunc is a registered trademark of Thermo Fisher Scientific or its subsidiaries
Nunclon is a registered trademark of Thermo Fisher Scientific or its subsidiaries
Ähnliche Artikel vergleichen
Vollständigen Vergleich anzeigen
Unterschiede anzeigen
1 of 1
Dieser Artikel
CLS430823CLS430639CLS430824
description
polystyrene, flask capacity 7 mL, Nunclon® Delta surface, sterile, case of 160 ea
description
surface area 150 cm2, canted neck, cap (plug seal)
description
surface area 25 cm2, canted neck, cap (vented)
description
surface area 150 cm2, canted neck, cap (phenolic-style)
manufacturer/tradename
Nunc 163371
manufacturer/tradename
Corning 430823
manufacturer/tradename
Corning 430639
manufacturer/tradename
Corning 430824
sterility
sterile
sterility
sterile
sterility
sterile
sterility
sterile
material
polystyrene
material
canted neck , rectangular flask, cap (plug seal), rectangular bottom flask, plug-seal screw-cap, polystyrene
material
rectangular flask, polystyrene , canted neck , vent cap, rectangular bottom flask, cap (vented)
material
phenolic cap, rectangular bottom flask, rectangular flask, cap (phenolic-style), canted neck , polystyrene
packaging
case of 160 ea
packaging
case of 50
packaging
case of 200
packaging
case of 50
flask capacity
7 mL
flask capacity
-
flask capacity
-
flask capacity
-
Hier finden Sie alle aktuellen Versionen:
Analysenzertifikate (COA)