Merck
TMO140652
Nunc® Cell Culture Inserts in 12-well Nunclon Delta surface plate
cell culture treated, polycarbonate, sterile, pore size 0.4 μm, case of 48 ea (supplied in 4 x 12 well plates)
Synonym(e):
multidish membrane insert, treated membrane insert, Nunc 140652, cultivation plate, surface treated dish
Anmelden zur Ansicht der Organisations- und Vertragspreise.
Größe auswählen
48 EA
€ 526,00
€ 526,00
Warenkorb auf Verfügbarkeit prüfen
Über diesen Artikel
NACRES:
NB.13
UNSPSC Code:
41122102
Material:
polycarbonate
Sterility:
sterile
Binding type:
cell culture treated
Manufacturer/tradename:
Nunc 140652
Packaging:
case of 48 ea (supplied in 4 x 12 well plates)
Fortfahren mit
Technischer Dienst
Benötigen Sie Hilfe? Unser Team von erfahrenen Wissenschaftlern ist für Sie da.
Unterstützung erhalten
material
polycarbonate
sterility
sterile
packaging
case of 48 ea (supplied in 4 x 12 well plates)
manufacturer/tradename
Nunc 140652
pore size
0.4 μm pore size
binding type
cell culture treated
Ähnliche Artikel vergleichen
Vollständigen Vergleich anzeigen
Unterschiede anzeigen
1 of 4
Dieser Artikel
TMO140654TMO140627TMO140656
sterility
sterile
sterility
sterile
sterility
sterile
sterility
sterile
material
polycarbonate
material
polycarbonate
material
polycarbonate
material
polycarbonate
manufacturer/tradename
Nunc 140652
manufacturer/tradename
Nunc 140654
manufacturer/tradename
Nunc 140627
manufacturer/tradename
Nalge-Nunc 140656
binding type
cell culture treated
binding type
cell culture treated
binding type
cell culture treated
binding type
cell culture treated
packaging
case of 48 ea (supplied in 4 x 12 well plates)
packaging
case of 48 ea (supplied in 4 x 12 well plates)
packaging
case of 48 ea (supplied as 4 x 24 well plates, each containing 12 inserts)
packaging
case of 48 ea (supplied in 4 x 12 well plates)
pore size
0.4 μm pore size
pore size
3.0 μm pore size
pore size
3.0 μm pore size
pore size
8.0 μm pore size
General description
Cultivate most cell types without matrix coating when using Thermo Scientific Nunc Polycarbonate Membrane Inserts in Multidishes.
Application
  • Tumor migration and invasion assays.
  • Epithelial (e.g. Caco-2) and endothelial (e.g. brain microvascular endothelial cells) barriers assays.
  • Dermal/epidermal and epithelial tissue models (e.g. artificial skin model).
  • Chemotaxis studies.
  • Co-culture.
Features and Benefits
  • Tissue culture-treated porous membrane for optimized cell attachment.
  • Three pore sizes: 0.4, 3, and 8 micron.
  • Pre-packed in sterile Nunclon® Delta-treated multi-well plate.s.
  • Nontoxic and nonpyrogenic.
Legal Information
Nunc is a registered trademark of Thermo Fisher Scientific or its subsidiaries
Hier finden Sie alle aktuellen Versionen:
Analysenzertifikate (COA)