Über diesen Artikel
NACRES:
NB.13
UNSPSC Code:
41122102
Material:
polycarbonate
Pore size:
8.0 μm pore size
Sterility:
sterile
Binding type:
cell culture treated
Technischer Dienst
Benötigen Sie Hilfe? Unser Team von erfahrenen Wissenschaftlern ist für Sie da.
Unterstützung erhaltenmaterial
polycarbonate
sterility
sterile
packaging
case of 48 ea (supplied in 4 x 12 well plates)
manufacturer/tradename
Nunc 140656
pore size
8.0 μm pore size
binding type
cell culture treated
General description
Cultivate most cell types without matrix coating when using Thermo Scientific Nunc Polycarbonate Membrane Inserts in Multidishes.
Application
- Tumor migration and invasion assays.
- Epithelial (e.g. Caco-2) and endothelial (e.g. brain microvascular endothelial cells) barriers assays.
- Dermal/epidermal and epithelial tissue models (e.g. artificial skin model).
- Chemotaxis studies.
- Co-culture.
Features and Benefits
- Tissue culture-treated porous membrane for optimized cell attachment.
- Three pore sizes: 0.4, 3, and 8 micron.
- Pre-packed in sterile Nunclon® Delta-treated multi-well plate.s.
- Nontoxic and nonpyrogenic.
Legal Information
Nunc is a registered trademark of Thermo Fisher Scientific or its subsidiaries
Still not finding the right product?
Explore all of our products under Nunc® Cell Culture Inserts in 12-well Nunclon Delta surface plate
Hier finden Sie alle aktuellen Versionen:
Besitzen Sie dieses Produkt bereits?