Ansicht ändern
| Größe/SKU | Verfügbarkeit | Preis |
|---|
Über diesen Artikel
NACRES:
NB.84
UNSPSC Code:
41122806
eCl@ss:
32032202
Manufacturer/tradename:
MTC™ Bio R2781-A
Feature:
to hold 81 x cryovials
Material:
assorted colors , cardboard
Packaging:
pkg of 5 ea
Preise und Verfügbarkeit sind derzeit nicht verfügbar.
Technischer Dienst
Benötigen Sie Hilfe? Unser Team von erfahrenen Wissenschaftlern ist für Sie da.
Unterstützung erhaltenmaterial
assorted colors , cardboard
feature
to hold 81 x cryovials
packaging
pkg of 5 ea
manufacturer/tradename
MTC™ Bio R2781-A
L × W × H
133 mm × 133 mm × 51 mm
General description
Recyclable cardboard, with a liquid resistant outer coating. The 81-place, 3 inch FlipTop™ box is suitable for tubes up to 13mm in diameter. The 100-place, 3 inch FlipTop™ box is suitable for tubes up to 12mm in diameter (accepts cryovials up to 4mL). Boxes are assembled complete with dividers. Each box has drain slots on the bottom and numbered grid markings in almost every conceivable location (top, bottom, sides).
Features and Benefits
- Attached Hinged Lids
- Foolproof identification
- 5 1/4 x 5 1/4 x 2 inches
- Standard size, for existing racks
- Available in 6 colors
- Individual or rainbow packs
- For 81 or 100 tubes
- 9×9 or 10×10 format
- Alpha-numeric coding
- Top, sides, and bottom
Legal Information
FlipTop is a trademark of MTC Bio
MTC is a trademark of MTC Bio
Still not finding the right product?
Explore all of our products under MTC™ Bio FlipTop™ Freezer Box With Hinged Lid
Hier finden Sie alle aktuellen Versionen:
Besitzen Sie dieses Produkt bereits?