Größe auswählen
| Ihnen/SKU | Verfügbarkeit | Preis |
|---|---|---|
1 ea | Warenkorb auf Verfügbarkeit prüfen | € 71,90 |
Über diesen Artikel
€ 71,90
material
acrylic , blue
feature
to hold 6 pipette(s)
packaging
pack of 1 ea
manufacturer/tradename
Heathrow Scientific HS20613C
L × W × H
295.3 mm × 172 mm × 203.2 mm
General description
Features and Benefits
- 6-Place Pipette Holder (1 in (25mm) Upper Slot, 0.8 in (20mm) Lower Slot)
- Designed to hold most major pipette brands
1 of 1
Dieser Artikel | |||
|---|---|---|---|
| description acrylic, to hold, 6 pipette(s), blue | description acrylic, to hold, 6 pipette(s), clear | description acrylonitrile-butadiene-styrene (ABS), to hold, 6 pipette(s), blue | description acrylic, to hold, 3 pipette(s), clear |
| feature to hold 6 pipette(s) | feature to hold 6 pipette(s) | feature to hold (pipettes), to hold 6 pipette(s) | feature to hold 3 pipette(s) |
| manufacturer/tradename Heathrow Scientific HS20613C | manufacturer/tradename Heathrow Scientific HS20620 | manufacturer/tradename Heathrow Scientific HS20613E | manufacturer/tradename Heathrow Scientific HS206203 |
| material acrylic , blue | material acrylic , clear | material acrylic , acrylonitrile-butadiene-styrene (ABS) , blue | material acrylic , clear |
| L × W × H 295.3 mm × 172 mm × 203.2 mm | L × W × H 295.3 mm × 172 mm × 203.2 mm | L × W × H 295.3 mm × 172 mm × 203.2 mm | L × W × H 153 mm × 172 mm × 206.4 mm |
| packaging pack of 1 ea | packaging pack of 1 ea | packaging pack of 1 ea | packaging pack of 1 ea |
Still not finding the right product?
Explore all of our products under Pipette Stand
Hier finden Sie alle aktuellen Versionen:
Besitzen Sie dieses Produkt bereits?