Größe auswählen
| Ihnen/SKU | Verfügbarkeit | Preis |
|---|
Über diesen Artikel
material
blue Nylon
feature
Holds 24 x 1.5-2 mL tubes
packaging
pack of 8 racks
manufacturer/tradename
Heathrow Scientific HS120241
technique(s)
PCR: suitable
well diam.
11.1 mm
Features and Benefits
- made from reinforced nylon
- superior chemical and heat resistance
- handles provide easy and safe transportation
- stackable two formats available for smaller and larger requirements
- temperature range -40 to 140C
- autoclavable
1 of 1
Dieser Artikel | |||
|---|---|---|---|
| description blue Nylon, pack of 8 racks, Holds 24 x 1.5-2 mL tubes | description blue Nylon, pack of 4 racks, Holds 96 x 1.5-2 mL tubes | description rectangular shaped, holds 24 tubes, Holds 24 x 1.5-2 mL tubes | description blue |
| feature Holds 24 x 1.5-2 mL tubes | feature Holds 96 x 1.5-2 mL tubes | feature Holds 24 x 1.5-2 mL tubes | feature Holds 12 x 15 mL tubes, Holds 32 x 0.5 mL tubes, Holds 32 x 1.5-2 mL tubes, Holds 4 x 50 mL tubes |
| manufacturer/tradename Heathrow Scientific HS120241 | manufacturer/tradename Heathrow Scientific HS120243 | manufacturer/tradename Heathrow Scientific HS2165C | manufacturer/tradename Heathrow Scientific HS29022B |
| material blue Nylon | material blue Nylon | material foamed PE , yellow | material polypropylene (heavy duty) |
| packaging pack of 8 racks | packaging pack of 4 racks | packaging - | packaging - |
| technique(s) PCR: suitable | technique(s) PCR: suitable | technique(s) - | technique(s) - |
| well diam. 11.1 mm | well diam. 11.1 mm | well diam. - | well diam. - |
Still not finding the right product?
Explore all of our products under Microtube Rack
Hier finden Sie alle aktuellen Versionen:
Besitzen Sie dieses Produkt bereits?