Merck
GBL629200
Grace Bio-Labs adhesive seal tabs for SecureSeal hybridization chambers
diam. 7.62 mm
Synonym(e):
adhesive seal tab, rubber seal tab
Größe auswählen
200 EA
€ 77,80
€ 77,80
Check Cart for Availability
Größe auswählen
Ansicht ändern
200 EA
€ 77,80
About This Item
UNSPSC-Code:
41122600
NACRES:
NB.22
€ 77,80
Check Cart for Availability
Sterilität
non-sterile
Verpackung
pack of 200 ea
Hersteller/Markenname
Grace Bio-Labs 629200
Durchmesser
7.62 mm
Wells
1
Allgemeine Beschreibung
Adhesive seal tabs complement SecureSeal Hybridization Chambers, HybriWell Sealing System, and CoverWell Perfusion Chambers. Once the paper liner is removed, the SecureSeal adhesive is exposed and the tab could easily be applied on top of a chamber’s port. A slight pressure on the adhesive tab is sufficient to create leak-proof chambers that are temperature resistant. Adhesive Seal Tabs are easily detached to allow addition or removal of reagents from the chamber. Adhesive Seal tabs are autoclavable.
Rechtliche Hinweise
CoverWell is a trademark of Grace Bio-Labs, Inc.
HybriWell is a trademark of Grace Bio-Labs, Inc.
SecureSeal is a trademark of Grace Bio-Labs, Inc.
Hier finden Sie alle aktuellen Versionen:
Analysenzertifikate (COA)