Merck
GBL112359
Grace Bio-Labs Culturewell chamberSLIP 16
wells, 16, with non-removable chambered coverglass, sterile, pack of 8
Synonym(e):
non-removable chambered coverglass, non-removable chambered coverslip
Größe auswählen
8 EA
€ 191,00
€ 191,00
Check Cart for Availability
Größe auswählen
Ansicht ändern
8 EA
€ 191,00
About This Item
UNSPSC-Code:
41161502
NACRES:
NB.17
€ 191,00
Check Cart for Availability
Sterilität
sterile
Leistungsmerkmale
wells 16
with non-removable chambered coverglass
Verpackung
pack of 8
Hersteller/Markenname
Grace Bio-Labs 112359
Well-Volumen × Fläche
100-250 μL × 0.4 cm2
Allgemeine Beschreibung
CultureWell ChamberSLIP 16 NON-Removable Chambered Coverglass, 16 Well, No. 1.5 German borosilicate coverglass. The product consists of cell culture vessels, with a 2×8 format with standard 96-well plate volumes and well spacing.
ChamberSLIP 16 Coverslip Non-Removable format has fixed chambers that do not separate from the coverglass for applications that do not require mounting fixed cells.
Rechtliche Hinweise
Culturewell is a trademark of Grace Bio-Labs, Inc.
Hier finden Sie alle aktuellen Versionen:
Analysenzertifikate (COA)
Questions
  1. What is the coverslip thickness? Are these #1.5 coverslips?
    1 answer
    1. Yes, the coverglass is #1.5, German borosilicate.