Größe auswählen
| Größe/SKU | Verfügbarkeit | Preis |
|---|---|---|
2 ea | Warenkorb auf Verfügbarkeit prüfen | € 335,00 |
Über diesen Artikel
€ 335,00
feature
Holds 9 mL tubes, fpr 17.5x100 mm round-bottom tubes
packaging
pack of 2 ea
manufacturer/tradename
Eppendorf® 5910762003
technique(s)
centrifugation: suitable
compatibility
for use with Universal bucket
Features and Benefits
- Broad range of adapters available for Rotor S-4xUniversal enables high versatility for tubes and flasks from 4 mL to 1,000 mL and plates
- Universal bucket design of Rotor S-4xUniversal accommodates both plates and tubes
- Universal adapters accommodate selective plate formats and conical tubes
- Aerosol-tight QuickLock caps for safe centrifugation of hazardous samples optionally available for Rotor S-4xUniversal
- Adapters are autoclavable
Legal Information
1 of 1
Dieser Artikel | |||
|---|---|---|---|
| description fpr 17.5x100 mm round-bottom tubes, pack of 2 ea | description 13x75-100 mm round-bottom rubes, pkg of 2 ea | description for 16x75-100 mm round-bottom tubes, pkg of 2 ea | description 0.5-2mL micro test tubes, rotor S-4xUniversal |
| manufacturer/tradename Eppendorf® 5910762003 | manufacturer/tradename Eppendorf® 5910755007 | manufacturer/tradename Eppendorf® 5910754000 | manufacturer/tradename 5910763000 |
| compatibility for use with Universal bucket | compatibility for use with Universal bucket | compatibility for use with Universal bucket | compatibility rotor S-4xUniversal |
| packaging pack of 2 ea | packaging pkg of 2 ea | packaging pkg of 2 ea | packaging pack of 2 ea |
| feature Holds 9 mL tubes, fpr 17.5x100 mm round-bottom tubes | feature Holds 4 - 8 mL tubes, 13x75-100 mm round-bottom rubes | feature Holds 7.5 - 12 mL tubes, for 16x75-100 mm round-bottom tubes | feature 0.5-2mL micro test tubes |
| technique(s) centrifugation: suitable | technique(s) centrifugation: suitable | technique(s) centrifugation: suitable | technique(s) - |
Still not finding the right product?
Explore all of our products under Adapter for Eppendorf® Rotor S-4xUniversal
Hier finden Sie alle aktuellen Versionen:
Besitzen Sie dieses Produkt bereits?