Merck
EP0030601904
Eppendorf® Microplate 96/V-PP, 96 wells
white polypropylene, conical bottom black wells, PCR clean, pkg of 80 ea
Größe auswählen
80 EA
€ 354,00
€ 354,00
Check Cart for Availability
Größe auswählen
Ansicht ändern
80 EA
€ 354,00
About This Item
UNSPSC-Code:
41122107
NACRES:
NB.15
€ 354,00
Check Cart for Availability
Materialien
conical bottom black wells
white polypropylene
Verpackung
pkg of 80 ea
Hersteller/Markenname
Eppendorf® 0030601904
Methode(n)
PCR: suitable
Größe
96 wells
Allgemeine Beschreibung
Pipette, control and recover your samples with ease using Eppendorf Microplates! Designed with the easy-to-read alphanumeric OptiTrack matrix label to identify individual wells, speed up the filling process and significantly reduce the risk of pipetting errors. Eppendorf Microplates are made with high-transparency polypropylene ensuring quick and easy location of samples
Rechtliche Hinweise
Eppendorf is a registered trademark of Eppendorf AG
Hier finden Sie alle aktuellen Versionen:
Analysenzertifikate (COA)