Größe auswählen
| Größe/SKU | Verfügbarkeit | Preis |
|---|---|---|
120 ea | Warenkorb auf Verfügbarkeit prüfen | € 248,00 |
Über diesen Artikel
€ 248,00
material
cap (plug seal), colorless cap, colorless tube, conical bottom
feature
PCR clean, graduations: no, neck thread Push fit, caps domed
packaging
pack of 120 ea
manufacturer/tradename
Eppendorf® 0030124928
technique(s)
PCR: suitable
capacity
0.1 mL
General description
- Ideal for use with Fast cyclers such as the Mastercycler X50s and Fast enzymes.
- 8 reaction tubes in strip-format − ideal for small sample volumes.
- Easily adaptable for automation.
- Sealable using domed strip-lids.
- Certified free from human DNA, DNase, RNase and PCR inhibitors.
Legal Information
1 of 1
Dieser Artikel | |||
|---|---|---|---|
| description caps domed, pack of 120 ea | description caps flat, pack of 120 ea | description capacity 0.1 mL, pack of 120 ea | description domed, pkg of 120 ea, 10 x 12 strips |
| manufacturer/tradename Eppendorf® 0030124928 | manufacturer/tradename Eppendorf® 0030124910 | manufacturer/tradename Eppendorf® 0030124901 | manufacturer/tradename Eppendorf® 0030124839 |
| capacity 0.1 mL | capacity 0.1 mL | capacity 0.1 mL | capacity - |
| material cap (plug seal), colorless tube, colorless cap, conical bottom | material conical bottom, cap (plug seal), colorless tube, colorless cap | material colorless tube, conical bottom, cap (plug seal) | material domed |
| packaging pack of 120 ea | packaging pack of 120 ea | packaging pack of 120 ea | packaging pkg of 120 ea |
| feature PCR clean, graduations: no, caps domed, neck thread Push fit | feature neck thread Push fit, graduations: no, PCR clean, caps flat | feature lid: no, PCR clean, | feature domed, autoclavable 120 °C |
| technique(s) PCR: suitable | technique(s) PCR: suitable | technique(s) PCR: suitable | technique(s) - |
Still not finding the right product?
Explore all of our products under Eppendorf® FAST PCR-Tube Strips
Hier finden Sie alle aktuellen Versionen:
Besitzen Sie dieses Produkt bereits?