Merck
CLS4519155
Corning® ProCulture® spinner flask accessories
Gas and Media Handling Fitting with Dual 1/4 in. Inlets for 3L Angled Sidearm Flasks
Synonym(e):
container accessory
Größe auswählen
1 EA
€ 576,00
€ 576,00
Check Cart for Availability
Größe auswählen
Ansicht ändern
1 EA
€ 576,00
About This Item
UNSPSC-Code:
41121804
NACRES:
NB.43
€ 576,00
Check Cart for Availability
Hersteller/Markenname
Corning 4519-155
Allgemeine Beschreibung
Side arm fittings used for a variety of functions in angled sidearm Spinner Flasks during cell culture. Sidearm fittings provide a convenient way of transferring media into or out of flasks aseptically. They can also be used to sparge the cell culture medium with gases to enhance cell production. Fittings comprise of a PET insert with a Viton O-ring and a polypropylene sealing cap & 316 stainless steel tubes.
Dual tubes extend to the bottom of the flask, and are held in place by Noryl nuts with integrated ferrules.
Combination tubes has two different length tubes, one extends to the bottom of the flask; the other is a shorter 6′ length. Both tubes are held in place by Noryl nuts with integrated ferrules.
Rechtliche Hinweise
Corning is a registered trademark of Corning, Inc.
ProCulture is a registered trademark of SP Industries
Hier finden Sie alle aktuellen Versionen:
Analysenzertifikate (COA)