Merck
CLS431846
Corning® centrifuge bottle
with silicon O-ring screw cap, capacity 500 mL, polycarbonate
Synonym(e):
centrifugation bottles, digestion bottles, shaking bottles
Größe auswählen
24 EA
€ 631,00
€ 631,00
Check Cart for Availability
Größe auswählen
Ansicht ändern
24 EA
€ 631,00
About This Item
UNSPSC-Code:
41121703
NACRES:
NB.11
€ 631,00
Check Cart for Availability
Materialien
clear bottle
flat bottom tube
polycarbonate
round bottle
wide-mouth bottle
Sterilität
non-sterile
Leistungsmerkmale
autoclavable
cap
Verpackung
pkg of 24 ea
Hersteller/Markenname
Corning 431846
Parameter
13700 × g max. RCF
Kapazität
500 mL
Leistungsmerkmale und Vorteile
  • Corning® centrifuge bottles have a specially designed cap for improved grip
  • Includes clear graduations and textured marking spot.
  • Bottles are designed with extra-thick walls for high durability
  • Wide mouth for easier liquid filling
  • Compatible with most centrifuges and do not require special buckets
  • Unique design aid researchers to reduce the number of centrifugation cycles and enhance productivity
Rechtliche Hinweise
Corning is a registered trademark of Corning, Inc.
Hier finden Sie alle aktuellen Versionen:
Analysenzertifikate (COA)